Skip to Content
Merck
All Photos(1)

Key Documents

SML1417

Sigma-Aldrich

Prosaptide Tx 14(A) trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

H-Thr-D-Ala-Leu-Ile-Asp-Asn-Asn-Ala-Thr-Glu-Glu-Ile-Leu-Tyr-OH trifluoroacetate salt, Prosaptide trifluoroacetate salt, TX14(A) trifluoroacetate salt, TXLIDNNATEEILY (Modifications: Ala-2 = D-Ala) trifluoroacetate salt, TaLIDNNATEEILY trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$289.00

$289.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$289.00

About This Item

Empirical Formula (Hill Notation):
C69H110N16O26 · xC2HF3O2
CAS Number:
Molecular Weight:
1579.70 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77

$289.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

N([C@@H]([C@H](O)C)C(=O)N[C@@H](CCC(=O)O)C(=O)N[C@@H](CCC(=O)O)C(=O)N[C@@H]([C@H](CC)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc1ccc(cc1)O)C(=O)O)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](NC(=O)[C@@H](N)[C@H](O)C)C

InChI

1S/C69H110N16O26/c1-13-31(7)53(66(107)80-41(23-29(3)4)61(102)82-46(69(110)111)25-37-15-17-38(88)18-16-37)83-59(100)40(20-22-50(93)94)75-58(99)39(19-21-49(91)92)76-68(109)55(36(12)87)85-57(98)34(10)73-60(101)43(26-47(70)89)78-62(103)44(27-48(71)90)79-63(104)45(28-51(95)96)81-67(108)54(32(8)14-2)84-64(105)42(24-30(5)6)77-56(97)33(9)74-65(106)52(72)35(11)86/h15-18,29-36,39-46,52-55,86-88H,13-14,19-28,72H2,1-12H3,(H2,70,89)(H2,71,90)(H,73,101)(H,74,106)(H,75,99)(H,76,109)(H,77,97)(H,78,103)(H,79,104)(H,80,107)(H,81,108)(H,82,102)(H,83,100)(H,84,105)(H,85,98)(H,91,92)(H,93,94)(H,95,96)(H,110,111)/t31-,32-,33+,34-,35+,36+,39-,40-,41-,42-,43-,44-,45-,46-,52-,53-,54-,55-/m0/s1

InChI key

SMUKRAODILXPSW-SRWCCNGVSA-N

Biochem/physiol Actions

Prosaptide Tx 14(A) is a potent agonist of G protein-coupled receptors GPR37 (also known as Parkin-associated endothelin receptor-like receptor Pael-R) and GPR37L1. Prosaptide Tx 14(A) is the active fragment of prosaposin, a secreted neuroprotective and glioprotective factor. Prosaptide Tx 14(A) has EC50 values of 5 nM for GPR37L1 and 7 nM for GPR37, and has also been shown to have some GSK3 inhibitory properties. Prosaptide has been shown to improve nerve regeneration in diabetic rats, to reduce neuropathic pain, and to protect dopaminergic neurons in animal models of Parkinson disease.
Prosaptide Tx 14(A) is a potent agonist of G protein-coupled receptors GPR37 and GPR37L1 with neuroprotective activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service