Skip to Content
Merck
All Photos(1)

Key Documents

SML1388

Sigma-Aldrich

N-Octanoyl dopamine

≥95% (HPLC)

Synonym(s):

N-(3,4-Dihydroxyphenethyl)octanamide, N-[2-(3,4-Dihydroxyphenyl)ethyl]-octanamide, NOD

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$136.00

$136.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$136.00

About This Item

Empirical Formula (Hill Notation):
C16H25NO3
CAS Number:
Molecular Weight:
279.37
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

$136.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=C(O)C=CC(CCNC(CCCCCCC)=O)=C1

InChI

1S/C16H25NO3/c1-2-3-4-5-6-7-16(20)17-11-10-13-8-9-14(18)15(19)12-13/h8-9,12,18-19H,2-7,10-11H2,1H3,(H,17,20)

InChI key

MGRVMTGNRVNIEL-UHFFFAOYSA-N

Biochem/physiol Actions

N-Octanoyl dopamine (NOD) protects cell cultures and tissues from cold storage inflicted damage.
N-Octanoyl dopamine (NOD) protects cell cultures and tissues from cold storage inflicted damage. N-Octanoyl dopamine decreases lactate-dehydrogenase (LDH) release during cold storage of cardiomyocytes in vitro. NOD did not induce a significant elevation of intracellular cAMP and is devoid of dopaminergic and adrenergic action.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service