Skip to Content
Merck
All Photos(1)

Key Documents

SML1346

Sigma-Aldrich

BRD32048

≥98% (HPLC)

Synonym(s):

BRD 32048, N2-(4-Methoxyphenyl)-6-(1-piperidinylmethyl)-1,3,5-triazine-2,4-diamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$155.00
25 MG
$496.00

$155.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$155.00
25 MG
$496.00

About This Item

Empirical Formula (Hill Notation):
C16H22N6O
CAS Number:
Molecular Weight:
314.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$155.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=NC(NC2=CC=C(OC)C=C2)=NC(CN3CCCCC3)=N1

InChI

1S/C16H22N6O/c1-23-13-7-5-12(6-8-13)18-16-20-14(19-15(17)21-16)11-22-9-3-2-4-10-22/h5-8H,2-4,9-11H2,1H3,(H3,17,18,19,20,21)

InChI key

PVHJDJWPRZUURH-UHFFFAOYSA-N

Biochem/physiol Actions

BRD32048 is a derivative of [1,3,5]triazine. It is found in orally active phosphodiesterase inhibitors (PDE) inhibitors, DHFR (dihydrofolate reductase) inhibitors and phosphatidylinositol-3-kinase (PI3K)/mammalian target of rapamycin (mTOR) inhibitors. BRD32048 binds ETS variant 1 (ETV1) and controls transcriptional activity, facilitated by ETV1 and invasion of ETV1-driven cancer cells.[1]
BRD32048 is a potent inhibitor of ETV1 (ETS variant 1) transcription factor oncoprotein.
BRD32048 is a potent inhibitor of ETV1 (ETS variant 1) transcription factor oncoprotein. BRD32048 binds ETV1 directly, modulating both ETV1-mediated transcriptional activity and invasion of ETV1-driven cancer cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A small molecule that binds and inhibits the ETV1 transcription factor oncoprotein
Pop MS, et al.
Molecular Cancer Therapeutics, molcanther-molcan0689 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service