Skip to Content
Merck
All Photos(1)

Key Documents

SML1332

Sigma-Aldrich

NG25 trihydrochloride

≥98% (HPLC)

Synonym(s):

N-[4-[(4-Ethyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-4-methyl-3-(1H-pyrrolo[2,3-b]pyridin-4-yloxy)-benzamide trihydrochloride, NG 25 trihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$185.00
25 MG
$741.00

$185.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$185.00
25 MG
$741.00

About This Item

Empirical Formula (Hill Notation):
C29H30F3N5O2 · 3HCl
CAS Number:
Molecular Weight:
646.96
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$185.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

H2O: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC=C(C(NC2=CC=C(CN3CCN(CC)CC3)C(C(F)(F)F)=C2)=O)C=C1OC4=C(C=CN5)C5=NC=C4

InChI

1S/C29H30F3N5O2/c1-3-36-12-14-37(15-13-36)18-21-6-7-22(17-24(21)29(30,31)32)35-28(38)20-5-4-19(2)26(16-20)39-25-9-11-34-27-23(25)8-10-33-27/h4-11,16-17H,3,12-15,18H2,1-2H3,(H,33,34)(H,35,38)

InChI key

SMPGEBOIKULBCT-UHFFFAOYSA-N

Application

NG25 trihydrochloride has been used as a transforming growth factor β (TGFβ)-activated kinase 1 inhibitor (TAK1)[1] in human umbilical vein endothelial cells (HUVECs)[2] and pulmonary artery smooth muscle cells.[3]

Biochem/physiol Actions

NG25 is multiple kinase inhibitor; potent dia; omjobotpr pf TAK1 and MAP4K2 kinases.
NG25 is potent multiple kinase inhibitor belonging to type II kinase inhibitors class that binds to kinase ATP binding pocket at not ready to catalyze conformation. NG25 is used as a potent dual inhibitor of TAK1 (TGFβ-Activated Kinase 1, MAP3K7) and MAP4K2 (Mitogen-Activated Protein Kinase Kinase Kinase Kinase 2; GCK) kinases. NG25 inhibits the activation of IKKa/IKKb and prevents the secretion of type 1 IFNs by TLR2 and TLR9 agonist.

Other Notes

Light sensitive

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliane Totzke et al.
Cell chemical biology, 24(8), 1029-1039 (2017-08-19)
Tumor necrosis factor alpha (TNF-α) has both positive and negative roles in human disease. In certain cancers, TNF-α is infused locally to promote tumor regression, but dose-limiting inflammatory effects limit broader utility. In autoimmune disease, anti-TNF-α antibodies control inflammation in
Lili Du et al.
American journal of physiology. Lung cellular and molecular physiology, 316(1), L20-L34 (2018-09-28)
TGFβ activation during newborn lung injury decreases the expression of pulmonary artery smooth muscle cell (PASMC)-soluble guanylate cyclase (sGC), a critical mediator of nitric oxide signaling. Using a rat PASMC line (CS54 cells), we determined how TGFβ downregulates sGC expression.
Ling-Yun Chu et al.
Scientific reports, 7(1), 12472-12472 (2017-10-01)
Pro-inflammatory cytokines are known to induce endothelial cell autophagy, but the role of autophagy in regulating the expression of pro-inflammatory molecules has not been characterized. We hypothesized that autophagy facilitates expression of endothelial adhesion molecules. TNFα and IL-1β induced autophagy
Yaron Shav-Tal et al.
FEBS letters, 531(2), 109-114 (2002-11-06)
Proteins are often referred to in accordance with the activity with which they were first associated or the organelle in which they were initially identified. However, a variety of nuclear factors act in multiple molecular reactions occurring simultaneously within the
Noam Fallik et al.
Oncotarget, 8(65), 109575-109586 (2018-01-10)
Hematopoietic Stem Cells (HSCs) generate blood and immune cells through a hierarchical process of differentiation. Genes that regulate this process are of great interest for understanding normal and also malignant hematopoiesis. Surprisingly, however, very little is known about long-non-coding RNAs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service