Skip to Content
Merck
All Photos(1)

Key Documents

SML1317

Sigma-Aldrich

XAP044

≥98% (HPLC)

Synonym(s):

7-Hydroxy-3-(4-iodophenoxy)-4H-chromen-4-one, XAP 044

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$621.00

$621.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$621.00

About This Item

Empirical Formula (Hill Notation):
C15H9IO4
CAS Number:
Molecular Weight:
380.13
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$621.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C(OC2=CC=C(I)C=C2)=COC3=C1C=CC(O)=C3

InChI

1S/C15H9IO4/c16-9-1-4-11(5-2-9)20-14-8-19-13-7-10(17)3-6-12(13)15(14)18/h1-8,17H

InChI key

XXZKJJYWINSUMJ-UHFFFAOYSA-N

Biochem/physiol Actions

XAP044 is a brain penetrant, potent and selective mGlu7 (metabotropic glutamate receptor subtype 7) antagonist that inhibits lateral amygdala long-term potentiation (LTP) in brain slices from wild type mice. XAP044 binds to a pocket localized in mGlu7′s extracellular Venus flytrap domain (VFTD) close to the binding site of the orthosteric agonist L-glutamate. XAP044 exhibits a wide-spectrum anti-stress, antidepressant- and anxiolytic-like efficacy in rodent behavioral model.
XAP044 is a brain penetrant, potent and selective mGlu7 antagonist.

Other Notes

Light sensitive. This compound has Iodine and should be kept in the dark.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service