Skip to Content
Merck
All Photos(1)

Key Documents

SML1310

Sigma-Aldrich

NSC756093

≥98% (HPLC)

Synonym(s):

4,9-Dihydro-4-(2-hydroxyethyl)-6-methoxy-9-phenyl-furo[3,4-b]quinolin-1(3H)-one, 4-(2-Hydroxyethyl)-6-methoxy-9-phenyl-4,9-dihydrofuro[3,4-b]quinolin-1(3H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$124.00
25 MG
$496.00

$124.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$124.00
25 MG
$496.00

About This Item

Empirical Formula (Hill Notation):
C20H19NO4
CAS Number:
Molecular Weight:
337.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$124.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

OCCN1C2=C(C(OC2)=O)C(C3=CC=CC=C3)C4=C1C=C(OC)C=C4

InChI

1S/C20H19NO4/c1-24-14-7-8-15-16(11-14)21(9-10-22)17-12-25-20(23)19(17)18(15)13-5-3-2-4-6-13/h2-8,11,18,22H,9-10,12H2,1H3

InChI key

SMEJQLRLHKWIFV-UHFFFAOYSA-N

Biochem/physiol Actions

NSC756093, a podophyllotoxin analog, is a potent and specific inhibitor of GBP1:PIM1 interaction that inhibits proliferation of paclitaxel resistant cancer cells. Apparently, NSC756093 stabilizes a conformation of GBP1 not suitable for binding to PIM1.
NSC756093, a podophyllotoxin analog, is a potent and specific inhibitor of GBP1:PIM1 interaction.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Zhao et al.
Frontiers in oncology, 9, 1494-1494 (2020-01-31)
The Guanylate binding proteins (GBPs) are a family of large GTPases and the most studied GBP family member is the guanylate binding protein 1 (GBP1). Earlier studies revealed that GBP1 expression was inflammatory cytokines-inducible, and most of the studies focused

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service