Skip to Content
Merck
All Photos(1)

Key Documents

SML1291

Sigma-Aldrich

Frovatriptan succinate monohydrate

≥97% (HPLC)

Synonym(s):

Frovatriptan succinate, R-(+) 3-Methylamino-6-carboxamido-1,2,3,4-tetrahydrocarbazole monosuccinate monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$155.00
50 MG
$496.00

$155.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$155.00
50 MG
$496.00

About This Item

Empirical Formula (Hill Notation):
C14H17N3O · C4H6O4 · H2O
CAS Number:
Molecular Weight:
379.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$155.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(CCC(O)=O)=O.NC(C(C=C1)=CC2=C1NC3=C2C[C@H](NC)CC3)=O.O

InChI

1S/C14H17N3O.C4H6O4.H2O/c1-16-9-3-5-13-11(7-9)10-6-8(14(15)18)2-4-12(10)17-13;5-3(6)1-2-4(7)8;/h2,4,6,9,16-17H,3,5,7H2,1H3,(H2,15,18);1-2H2,(H,5,6)(H,7,8);1H2/t9-;;/m1../s1

InChI key

CUETXFMONOSVJA-KLQYNRQASA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

During migraine condition, frovatriptan succinate monohydrate acts on extra cerebral, intracranial arteries and prevent excessive dilation of these vessels.[1]
Frovatriptan is a serotonin 5HT-1B/1D-receptor agonist used to treat migraine. Frovatriptan has high affinity for the 5-HT(1B) and 5-HT(1D) receptors while affinity for 5-HT receptors other than 5-HT(1B/1D) is substantially lower with a pEC50 value of 8.2 for the 5-HT1B receptor, compared to a pEC50 value of 6.2 for the 5-HT7 receptor.
Frovatriptan is a serotonin 5HT-1B/1D-receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple Spectrophotometric Method for Estimation of Frovatriptan Succinate in Bulk Drug and Pharmaceutical Formulation.
Nayak S, et al.
International Journal of Pharmaceutical Research and Allied Sciences, 4(2), 85-89 (2015)
Barbora Vyhlídalová et al.
International journal of molecular sciences, 21(8) (2020-04-23)
The efforts for therapeutic targeting of the aryl hydrocarbon receptor (AhR) have emerged in recent years. We investigated the effects of available antimigraine triptan drugs, having an indole core in their structure, on AhR signaling in human hepatic and intestinal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service