Skip to Content
Merck
All Photos(1)

Key Documents

SML1254

Sigma-Aldrich

R547

≥98% (HPLC)

Synonym(s):

Ro 4584820, [4-Amino-2-(1-methanesulfonylpiperidin-4-ylamino)pyrimidin-5-yl](2,3-difluoro-6-methoxyphenyl)methanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$141.00

$141.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$141.00

About This Item

Empirical Formula (Hill Notation):
C18H21F2N5O4S
CAS Number:
Molecular Weight:
441.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$141.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

NC1=NC(NC2CCN(S(=O)(C)=O)CC2)=NC=C1C(C3=C(OC)C=CC(F)=C3F)=O

InChI

1S/C18H21F2N5O4S/c1-29-13-4-3-12(19)15(20)14(13)16(26)11-9-22-18(24-17(11)21)23-10-5-7-25(8-6-10)30(2,27)28/h3-4,9-10H,5-8H2,1-2H3,(H3,21,22,23,24)

InChI key

JRNJNYBQQYBCLE-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

R547 is a potent and selective ATP-competitive inhibitor of Cyclin-Dependent Kinases (CDK) 1, 2 and 4 with Ki of 2 nM for CDK1/cyclin B, 3 nM for CDK2/cyclin E, and 1 nM for CDK4/cyclin D1. R547 is less potent for CDK7 and GSK3α/β, and inactive (Ki > 5,000 nmol/L) against a panel of >120 unrelated kinases. R547 was shown to inhibit retinoblastoma protein in tumor cells and xenografts and showed up to 95% tumor growth inhibition in the HCT116 human colorectal tumor xenograft model in nude mice.
R547 is a potent and selective ATP-competitive inhibitor of Cyclin-Dependent Kinases (CDK) 1, 2 and 4.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehdi Bouhaddou et al.
Cell, 182(3), 685-712 (2020-07-10)
The causative agent of the coronavirus disease 2019 (COVID-19) pandemic, severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has infected millions and killed hundreds of thousands of people worldwide, highlighting an urgent need to develop antiviral therapies. Here we present a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service