Skip to Content
Merck
All Photos(1)

Key Documents

SML1237

Sigma-Aldrich

CCT137690

≥98% (HPLC)

Synonym(s):

6-Bromo-7-[4-[(5-methyl-3-isoxazolyl)methyl]-1-piperazinyl]-2-[4-(4-methyl-1-piperazinyl)phenyl]-3H-imidazo[4,5-b]pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$156.00

$156.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$156.00

About This Item

Empirical Formula (Hill Notation):
C26H31BrN8O
CAS Number:
Molecular Weight:
551.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$156.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

CN(CC1)CCN1C(C=C2)=CC=C2C3=NC4=C(N5CCN(CC6=NOC(C)=C6)CC5)C(Br)=CN=C4N3

InChI

1S/C26H31BrN8O/c1-18-15-20(31-36-18)17-33-9-13-35(14-10-33)24-22(27)16-28-26-23(24)29-25(30-26)19-3-5-21(6-4-19)34-11-7-32(2)8-12-34/h3-6,15-16H,7-14,17H2,1-2H3,(H,28,29,30)

InChI key

GFLQCBTXTRCREJ-UHFFFAOYSA-N

Biochem/physiol Actions

CCT137690 is a very potent, orally bioavailble, pan aurora kinase inhbitor (IC50 = 15, 25, and 19 nM for Aurora A, B and C respectively). CCT137690 inhibits tumor growth in amouse xenograft model and causes multipolar spindle formation, chromosome misalignment, polyploidy and apoptosis in tumor cell lines.
CCT137690 is a very potent, orally bioavailble, pan aurora kinase inhbitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service