Skip to Content
Merck
All Photos(1)

Key Documents

SML1191

Sigma-Aldrich

CX516

≥98% (HPLC)

Synonym(s):

1-Piperidinyl-6-quinoxalinyl-methanone, 6-(Piperidin-1-ylcarbonyl)quinoxaline, BDP-12;, CX-516

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$141.00
25 MG
$567.00

$141.00


Estimated to ship on03 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$141.00
25 MG
$567.00

About This Item

Empirical Formula (Hill Notation):
C14H15N3O
CAS Number:
Molecular Weight:
241.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$141.00


Estimated to ship on03 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

H2O: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC2=NC=CN=C2C=C1)N3CCCCC3

InChI

1S/C14H15N3O/c18-14(17-8-2-1-3-9-17)11-4-5-12-13(10-11)16-7-6-15-12/h4-7,10H,1-3,8-9H2

InChI key

ANDGGVOPIJEHOF-UHFFFAOYSA-N

Biochem/physiol Actions

CX516 is a positive allosteric modulator at AMPA receptor that inhibits the deactivation of AMPA receptors. CX-516 is a nootropic and ampakine agent.
CX516 is a positive allosteric modulator at AMPA receptor; cognitive enhancer.

Other Notes

Air sensitive

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service