Skip to Content
Merck
All Photos(1)

Key Documents

SML1187

Sigma-Aldrich

Vacquinol-1 dihydrochloride

≥98% (HPLC)

Synonym(s):

2-(4-Chlorophenyl)-α−2-piperidinyl-4-quinolinemethanol dihydrochloride, NSC13316

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$150.00

$150.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$150.00

About This Item

Empirical Formula (Hill Notation):
C21H21ClN2O · 2HCl
CAS Number:
Molecular Weight:
425.78
UNSPSC Code:
12352200
NACRES:
NA.77

$150.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)c2nc3c(c(c2)C(O)C4NCCCC4)cccc3

InChI

1S/C21H21ClN2O/c22-15-10-8-14(9-11-15)20-13-17(16-5-1-2-6-18(16)24-20)21(25)19-7-3-4-12-23-19/h1-2,5-6,8-11,13,19,21,23,25H,3-4,7,12H2

InChI key

VKLJPGAHSLIQKH-UHFFFAOYSA-N

Biochem/physiol Actions

Vacquinol-1 (NSC13316) is a brain penetrant inducer of catastrophic vacuolization in glioblastoma cells (GCs). Vacquinol-1 displays high cytotoxicity to GCs; it induces cell death by membrane ruffling, cell rounding, massive macropinocytic vacuole accumulation, ATP depletion, and cytoplasmic membrane rupture of GCs. Vacquinol-1 originally was developed as an antimalarial compound.
Vacquinol-1 dihydrochloride is potentially used as a therapeutic for Glioblastoma multiforme (GBM). This drug is highly stable in vivo, with a half-life in plasma of 52 hour. Vacquinol-1 has high oral bioavailability and good brain penetration.[1]
Vacquinol-1 is a brain penetrant inducer of catastrophic vacuolization in glioblastoma cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Driving glioblastoma to drink.
Gilbertson R J.
Cell, 157(2), 289-290 (2014)
Jie Zhen et al.
The American journal of Chinese medicine, 49(1), 147-161 (2020-12-30)
Acute pancreatitis (AP) is a kind of reversible inflammatory process of the exocrine pancreas. During the process, systemic inflammatory syndromes are involved, which relates closely to inflammatory mediators. Baicalin is a type of flavone compound extracted from Scutellaria baicalensis Georgi
Gi Bum Kim et al.
Cell, 179(1), 251-267 (2019-09-21)
In situ transgenesis methods such as viruses and electroporation can rapidly create somatic transgenic mice but lack control over copy number, zygosity, and locus specificity. Here we establish mosaic analysis by dual recombinase-mediated cassette exchange (MADR), which permits stable labeling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service