Skip to Content
Merck
All Photos(1)

Key Documents

SML1172

Sigma-Aldrich

FFN102

≥98% (HPLC)

Synonym(s):

4-(2-Amino-ethyl)-6-chloro-7-hydroxy-chromen-2-one trifluoroacetate, 4-(2-Aminoethyl)-6-chloro-7-hydroxy-2H-1-benzopyran-2-one trifluoroacetate, Mini 102, Mini102

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$209.00

$209.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$209.00

About This Item

Empirical Formula (Hill Notation):
C11H10ClNO3 · CF3CO2H
CAS Number:
Molecular Weight:
353.68
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$209.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)C(O)=O.NCCC(C1=CC(Cl)=C(O)C=C1O2)=CC2=O

InChI

1S/C11H10ClNO3.C2HF3O2/c12-8-4-7-6(1-2-13)3-11(15)16-10(7)5-9(8)14;3-2(4,5)1(6)7/h3-5,14H,1-2,13H2;(H,6,7)

InChI key

OGHPXKAKVMJGQH-UHFFFAOYSA-N

Biochem/physiol Actions

FFN102 is a pH-responsive fluorescent false neurotransmitter that acts as a dopamine transporter substrate (DAT) and also a substrate for vesicular monoamine transporter 2 (VMAT2).
FFN102 is a pH-responsive fluorescent false neurotransmitter that acts as a dopamine transporter substrate (DAT) and also a substrate for vesicular monoamine transporter 2 (VMAT2). FFN102 selectively labels dopamine cell bodies and dendrites in ventral midbrain and dopaminergic synaptic terminals in dorsal striatum. Its greater fluorescence emission in neutral than acidic environments allows the visualization of dopamine release at individual presynaptic terminals and in situ pH measurement of catecholamine secretory vesicles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service