Skip to Content
Merck
All Photos(1)

Key Documents

SML1142

Sigma-Aldrich

VU0071063

≥98% (HPLC)

Synonym(s):

7-[[4-(1,1-Dimethylethyl)phenyl]methyl]-3,7-dihydro-1,3-dimethyl-1H-purine-2,6-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$142.00
25 MG
$450.00

$142.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$142.00
25 MG
$450.00

About This Item

Empirical Formula (Hill Notation):
C18H22N4O2
CAS Number:
Molecular Weight:
326.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$142.00


Estimated to ship on25 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C1=C2N(CC3=CC=C(C(C)(C)C)C=C3)C=N1)C(N(C)C2=O)=O

InChI

1S/C18H22N4O2/c1-18(2,3)13-8-6-12(7-9-13)10-22-11-19-15-14(22)16(23)21(5)17(24)20(15)4/h6-9,11H,10H2,1-5H3

InChI key

ZFZAIHKQJBMYLO-UHFFFAOYSA-N

Biochem/physiol Actions

VU0071063 is a potent activator of Kir6.2/SUR1 KATP channels (EC50 = 10.3 μM). VU0071063 reversibly inhibits glucose-stimulated calcium influx in mouse β-islet cells. The compound VU0071063 has no affect on Kir6.2/SUR2A channels.
VU0071063 is a specific activator of SUR1-containing KATP channels.
VU0071063 is also known as [7-(4-(tert-butyl)benzyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione]. It serves as a major compound for examining β-cell physiology, KATP channel gating and a new chemical scaffold for evolving enhanced activators with medicinal chemistry.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct activation of beta-cell KATP channels with a novel xanthine derivative
Raphemot RS, et al.
Molecular Pharmacology, mol-114 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service