Skip to Content
Merck
All Photos(1)

Key Documents

SML1119

Sigma-Aldrich

LDN-214117

≥98% (HPLC)

Synonym(s):

1-(4-(6-Methyl-5-(3,4,5-trimethoxyphenyl)pyridin-3-yl)phenyl)piperazine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$250.00
25 MG
$661.00

$250.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$250.00
25 MG
$661.00

About This Item

Empirical Formula (Hill Notation):
C25H29N3O3
CAS Number:
Molecular Weight:
419.52
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$250.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C(C1=CC(OC)=C(OC)C(OC)=C1)=C2)=NC=C2C(C=C3)=CC=C3N4CCNCC4

InChI

1S/C25H29N3O3/c1-17-22(19-14-23(29-2)25(31-4)24(15-19)30-3)13-20(16-27-17)18-5-7-21(8-6-18)28-11-9-26-10-12-28/h5-8,13-16,26H,9-12H2,1-4H3

InChI key

BHUXVRVMMYAXKN-UHFFFAOYSA-N

Application

LDN-214117 has been used in structure-activity relationship (SAR) study to evaluate its potency on selective inhibition of activin receptor-like kinase-2 (ALK2) through the kinome-wide selectivity of (LDN-214117) via enzymatic kinase profiling.[1]

Biochem/physiol Actions

LDN-214117 is a selective inhibitor of the bone morphogenetic protein (BMP) type I receptor kinases with high selectivity for BMP versus TGF-β signaling, and low cytotoxicity. LDN-214117 inhibited ALK2 most, with a biochemical IC50 of 24 nM, followed by TNIK, RIPK2, and ABL1. LDN-214117 has a cell-based IC50 for BMP6 of approximately 100 nM and 164-fold selectivity for BMP6 versus TGF-β1. Fibrodysplasia ossificans progressiva (FOP) is a debilitating and progressive heterotopic ossification disease caused by activating mutations of ACVR1 encoding the BMP type I receptor kinase ALK2. LDN-214117 had nearly identical binding affinity for wild-type ALK2 and each of the FOP-causing mutants tested.
LDN-214117 is a selective inhibitor of the bone morphogenetic protein (BMP) type I receptor kinases.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agustin H Mohedas et al.
Journal of medicinal chemistry, 57(19), 7900-7915 (2014-08-08)
There are currently no effective therapies for fibrodysplasia ossificans progressiva (FOP), a debilitating and progressive heterotopic ossification disease caused by activating mutations of ACVR1 encoding the BMP type I receptor kinase ALK2. Recently, a subset of these same mutations of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service