Skip to Content
Merck
All Photos(1)

Key Documents

SML1011

Sigma-Aldrich

INDY

≥98% (HPLC)

Synonym(s):

(1Z)-1-(3-Ethyl-5-hydroxy-2(3H)-benzothiazolylidene)-2-propanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$99.30
25 MG
$395.00

$99.30


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$99.30
25 MG
$395.00

About This Item

Empirical Formula (Hill Notation):
C12H13NO2S
CAS Number:
Molecular Weight:
235.30
UNSPSC Code:
12352200
NACRES:
NA.77

$99.30


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

S1c2c(cc(cc2)O)N(\C\1=C/C(=O)C)CC

InChI

1S/C12H13NO2S/c1-3-13-10-7-9(15)4-5-11(10)16-12(13)6-8(2)14/h4-7,15H,3H2,1-2H3/b12-6+

InChI key

GCSZJMUFYOAHFY-WUXMJOGZSA-N

Biochem/physiol Actions

INDY is a potent inhibitor of Dyrk1A and Dyrk1B (dual-specificity tyrosine-phosphorylation-regulated kinase 1). The IC50 values for inhibition of Dyrk1A and 1B are 240 and 230 nM, respectively. INDY impairs the self-renewal capacity of neural stem cells and glioblastoma tumor-initiating cells.
INDY is a potent inhibitor of Dyrk1A and Dyrk1B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service