Skip to Content
Merck
All Photos(1)

Key Documents

SML0916

Sigma-Aldrich

CaCCinh-A01

≥98% (HPLC)

Synonym(s):

6-(1,1-Dimethylethyl)-2-[(2-furanylcarbonyl)amino]-4,5,6,7-tetrahydro-benzo[b]thiophene-3-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$167.00
25 MG
$449.00

$167.00


Available to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$167.00
25 MG
$449.00

About This Item

Empirical Formula (Hill Notation):
C18H21NO4S
CAS Number:
Molecular Weight:
347.43
UNSPSC Code:
12352200
NACRES:
NA.77

$167.00


Available to ship on09 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c2c(c(c1NC(=O)c3[o]ccc3)C(=O)O)CCC(C2)C(C)(C)C

InChI

1S/C18H21NO4S/c1-18(2,3)10-6-7-11-13(9-10)24-16(14(11)17(21)22)19-15(20)12-5-4-8-23-12/h4-5,8,10H,6-7,9H2,1-3H3,(H,19,20)(H,21,22)

InChI key

ACLUEOBQFRYTQS-UHFFFAOYSA-N

Biochem/physiol Actions

CaCCinh-A01 is a TMEM16A (transmembrane member 16A) inhibitor.[1] It also blocks anoctamin-1 (ANO1) and decreases cell viability and represses cell migration.[2]
CaCCinh-A01 is a potent calcium-activated chloride channel (CaCC) inhibitor that fully inhibits CaCC current in human bronchial and intestinal cells. CaCCinh-A01 does not inhibit CFTR.
CaCCinh-A01 is a potent calcium-activated chloride channel (CaCC) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Inhibition of calcium-activated chloride channel ANO1 suppresses proliferation and induces apoptosis of epithelium originated cancer cells.
Guan L, et al.
Oncotarget, 7(48), 78619-78619 (2016)
Ca2+-activated Cl? channel currents in mammary secretory cells from lactating mouse.
Kamikawa A, et al.
American Journal of Physiology. Cell Physiology, 311(5), C808-C819 (2016)
Ana N Gonçalves et al.
Respiratory research, 24(1), 42-42 (2023-02-06)
Clinical and experimental evidence shows lung fluid volume as a modulator of fetal lung growth with important value in treating fetal lung hypoplasia. Thus, understanding the mechanisms underlying these morphological dynamics has been the topic of multiple investigations with, however
Hayley Pearson et al.
Thorax, 76(1), 64-72 (2020-10-29)
Human respiratory syncytial virus (HRSV) is a common cause of respiratory tract infections (RTIs) globally and is one of the most fatal infectious diseases for infants in developing countries. Of those infected, 25%-40% aged ≤1 year develop severe lower RTIs leading to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service