Skip to Content
Merck
All Photos(1)

Key Documents

SML0904

Sigma-Aldrich

ABO dihydrochloride

≥98% (HPLC)

Synonym(s):

6-Amino-2, 3-dihydro-3-hydroxymethyl-1, 4-benzoxazine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$96.90
25 MG
$382.00

$96.90


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$96.90
25 MG
$382.00

About This Item

Empirical Formula (Hill Notation):
C9H12N2O2 · 2HCl
CAS Number:
Molecular Weight:
253.13
UNSPSC Code:
41106300
NACRES:
NA.77

$96.90


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

, light yellow to grey to brown

solubility

H2O: 25 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

ABO is a modulator of Annexin A7 (ANXA7) GTPase, a member of the annexin family of calcium/phospholipid-binding proteins. ABO can directly bind to Annexin A7 and inhibit its phosphorylation. Annexin A7 is involved in several processes especially in the process of membrane fusion and exocytosis. ABO is a GTPase, and both GTP-binding and Protein kinase C (PKC) activity are important in regulating its function to potentiate calcium-dependent membrane fusion. Annexin A7 has several other activities, not fully understood. ABO suppressed oxidized low-density lipoprotein (oxLDL)-induced phosphatidylcholine-specific phospholipase C (PC-PLC) activity, inhibiting apoptosis and promoting autophagy in vascular endothelial cells, indicating that ANXA7 is an endogenous regulator of phosphatidylcholine-specific phospholipase C (PC-PLC). ABO significantly reduced atherosclerotic plaque area and reduced lipid deposition and pro-inflammatory macrophages in apolipoprotein E-/- mice. ABO should be a valuable tool to study the complex functions of Annexin 7.
ABO is a modulator of Annexin A7 (ANXA7) GTPase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service