Skip to Content
Merck
All Photos(1)

Key Documents

SML0883

Sigma-Aldrich

HIF-2 Antagonist 2

≥98% (HPLC)

Synonym(s):

N-(3-Chloro-5-fluorophenyl)-4-nitrobenzo[c][1,2,5]oxadiazol-5-amine, TC-S7009

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$142.00
25 MG
$382.00

$142.00


Estimated to ship on21 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$142.00
25 MG
$382.00

About This Item

Empirical Formula (Hill Notation):
C12H6ClFN4O3
CAS Number:
Molecular Weight:
308.65
UNSPSC Code:
12352200
NACRES:
NA.77

$142.00


Estimated to ship on21 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

room temp

SMILES string

Fc1cc(cc(c1)Nc2c(c3n[o]nc3cc2)[N+](=O)[O-])Cl

InChI

1S/C12H6ClFN4O3/c13-6-3-7(14)5-8(4-6)15-10-2-1-9-11(17-21-16-9)12(10)18(19)20/h1-5,15H

InChI key

CDQUJZKBRAFWNG-UHFFFAOYSA-N

Biochem/physiol Actions

HIF-2α antagonist.
Hypoxia inducible transcription factors (HIF) control gene expression when oxygen availability is low and are associated with the onset and progression of a variety of cancers. They are heterodimers of an HIF-α (HIF-1α, HIF-2α also known as EPAS-1, or HIF-3α) and aryl hydrocarbon receptor nuclear translocator (ARNT, also known as HIF-β). N-(3-Chloro-5-fluorophenyl)-4-nitrobenzo[c][1,2,5]oxadiazol-5-amine is a highly selective antagonist of HIF-2 heterodimerization, DNA-binding activity, and target gene transcription. It binds the HIF-2α PAS-B domain with a KD ~80 nM compared to a KD > 5000 nM for the HIF-1α PAS-B domain.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabelle Westerlund et al.
Biochemical and biophysical research communications, 508(4), 1233-1239 (2018-12-20)
The hypoxia inducible transcription factor EPAS1/HIF2α has been described as an oncogene and a potential therapeutic target in neuroblastoma. Our analysis of several neuroblastoma tumour expression datasets does not support an oncogenic role, instead EPAS1 expression is associated with better
Kyoko Endo et al.
International journal of molecular sciences, 21(1) (2019-12-22)
Previous studies have reported the up-regulation of the two-pore domain K+ channel K2P5.1 in the CD4+ T cells of patients with multiple sclerosis (MS) and rheumatoid arthritis (RA), as well as in a mouse model of inflammatory bowel disease (IBD).
Bo Zhang et al.
Cells, 11(16) (2022-08-27)
Hypoxia-inducible factor (HIF) directly activates the transcription of metabolic enzymes in response to hypoxia to reprogram cellular metabolism required for tumor cell proliferation. Through analyzing glutamate-linked aminotransferases, we here identified glutamate pyruvate transaminase 2 (GPT2) as a direct HIF-2 target
Fumihito Hikage et al.
Endocrinology, 160(1), 20-35 (2018-11-06)
Thyroid-associated orbitopathy (TAO) is a disfiguring periocular connective tissue disease associated with autoimmune thyroid disorders. It is a potentially blinding condition, for which no effective pharmacological treatment has been established. Despite a suggested role played by autoimmune thyrotropin receptor activation

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service