Skip to Content
Merck
All Photos(1)

Key Documents

SML0835

Sigma-Aldrich

PF74

≥98% (HPLC)

Synonym(s):

2-Methyl-N-[(1S)-2-(methylphenylamino)-2-oxo-1-(phenylmethyl)ethyl]- 1H-indole-3-acetamide, PF 3450074, PF-3450074

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$177.00
25 MG
$450.00

$177.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$177.00
25 MG
$450.00

About This Item

Empirical Formula (Hill Notation):
C27H27N3O2
CAS Number:
Molecular Weight:
425.52
UNSPSC Code:
12352200
NACRES:
NA.77

$177.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[nH]1c2c(c(c1C)CC(=O)N[C@@H](Cc4ccccc4)C(=O)N(C)c3ccccc3)cccc2

InChI

1S/C27H27N3O2/c1-19-23(22-15-9-10-16-24(22)28-19)18-26(31)29-25(17-20-11-5-3-6-12-20)27(32)30(2)21-13-7-4-8-14-21/h3-16,25,28H,17-18H2,1-2H3,(H,29,31)/t25-/m0/s1

InChI key

ACDFWSNAQWFRRF-VWLOTQADSA-N

Biochem/physiol Actions

PF74 (PF-3450074) is a HIV-1 inhibitor that targets HIV capsid protein.
PF74 (PF-3450074) is a HIV-1 inhibitor that targets HIV capsid protein. PF74 binds specifically to HIV-1 particles and triggers premature HIV-1 uncoating in target cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D S Heller et al.
International journal of fertility, 37(5), 283-285 (1992-09-01)
Patients with complete androgen insensitivity syndrome rarely have Müllerian remnants. A patient with this syndrome found to have a microscopic fallopian tube at the time of gonadectomy is described and possible etiologies for the finding are discussed.
Muthukumar Balasubramaniam et al.
Journal of virology, 93(6) (2018-12-21)
The HIV-1 capsid protein (CA) facilitates reverse transcription and nuclear entry of the virus. However, CA's role in post-nuclear entry steps remains speculative. We describe a direct link between CA and integration by employing the capsid inhibitor PF74 as a
Doortje Borrenberghs et al.
Scientific reports, 6, 36485-36485 (2016-11-11)
Nuclear entry is a selective, dynamic process granting the HIV-1 pre-integration complex (PIC) access to the chromatin. Classical analysis of nuclear entry of heterogeneous viral particles only yields averaged information. We now have employed single-virus fluorescence methods to follow the
Muthukumar Balasubramaniam et al.
Journal of virology, 93(6) (2018-12-21)
The HIV-1 capsid protein (CA) facilitates reverse transcription and nuclear entry of the virus. However, CA's role in post-nuclear entry steps remains speculative. We describe a direct link between CA and integration by employing the capsid inhibitor PF74 as a
Alžběta Dostálková et al.
Molecules (Basel, Switzerland), 25(8) (2020-04-25)
A major structural retroviral protein, capsid protein (CA), is able to oligomerize into two different hexameric lattices, which makes this protein a key component for both the early and late stages of HIV-1 replication. During the late stage, the CA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service