Skip to Content
Merck
All Photos(1)

Key Documents

SML0806

Sigma-Aldrich

7DG

≥95% (HPLC)

Synonym(s):

7-Desacetoxy-6,7-dehydrogedunin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$69.00
5 MG
$283.00

$69.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
1 MG
$69.00
5 MG
$283.00

About This Item

Empirical Formula (Hill Notation):
C26H30O5
CAS Number:
Molecular Weight:
422.51
UNSPSC Code:
12352200
NACRES:
NA.77

$69.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

shipped in

wet ice

storage temp.

2-8°C

General description

7-Desacetoxy-6,7-dehydrogedunin (7DG) is structurally similar to HSP90 (heat shock protein 90) inhibitor, also known as gedunin.[1]

Biochem/physiol Actions

7-Desacetoxy-6,7-dehydrogedunin (7DG) is a selective inhibitor of protein kinase R (PKR).
7-Desacetoxy-6,7-dehydrogedunin (7DG) is a selective inhibitor of protein kinase R (PKR). 7DG appears to directly interact with the C-terminal half of PKR, and unlike C16, does not bind the ATP catalytic pocket. 7-Desacetoxy-6,7-dehydrogedunin completely protected macrophages from anthrax lethal toxin (LT)-induced pyroptotic cell death, a model for inflammasome-mediated caspase-1 activation, with an (IC50) of 5 μM, showing a role for PKR in pyroptosis.

Features and Benefits

This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik C Hett et al.
Nature chemical biology, 9(6), 398-405 (2013-04-23)
Formation of the inflammasome, a scaffolding complex that activates caspase-1, is important in numerous diseases. Pyroptotic cell death induced by anthrax lethal toxin (LT) is a model for inflammasome-mediated caspase-1 activation. We discovered 7-desacetoxy-6,7-dehydrogedunin (7DG) in a phenotypic screen as

Articles

Agents reported to activate cellular caspases include chemotherapeutic drugs, TNF receptor agonists, and other enzymes. Inhibitors of apoptosis were the first identified endogenous caspase inhibitors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service