Skip to Content
Merck
All Photos(1)

Documents

SML0774

Sigma-Aldrich

SR 3677

≥98% (HPLC)

Synonym(s):

N-[2-[2-(Dimethylamino)ethoxy]-4-(1H-pyrazol-4-yl)phenyl]-2,3-dihydro-1,4-benzodioxin-2-carboxamide., SR-3677, SR3677

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N4O4
CAS Number:
Molecular Weight:
408.45
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C22H24N4O4/c1-26(2)9-10-28-20-11-15(16-12-23-24-13-16)7-8-17(20)25-22(27)21-14-29-18-5-3-4-6-19(18)30-21/h3-8,11-13,21H,9-10,14H2,1-2H3,(H,23,24)(H,25,27)

InChI key

OQWZIAVXCYIZNN-UHFFFAOYSA-N

Application

SR 3677 has been used as Rho-associated kinase (ROCK) inhibitor in cardiac reprogramming.

Biochem/physiol Actions

SR-3677 is a very potent, selective ROCK inhibitor, with higher potency against ROCK II (IC50 = 3 nM) than ROCK I (IC50 = 56 nM). In a kinase screening panel, SR-3677 was shown to only have activity against three other kinases (PKA, MRCK and Akt1) all with IC50 values over 1 μM. The compound has nM potency in cell based assays, and is efficacious in an ex-vivo model for increasing aqueous humor outflow in porcine eyes.
SR3677 decreases amyloid precursor protein (sAPPβ) and amyloid-β (Aβ) in a dose-dependent manner. In 5XFAD mouse brain, SR3677 represses the enzymatic activity of β-site APP cleaving enzyme 1 (BACE1). SR3677 has the ability to change the endocytic supply of BACE1 and induce APP traffic to lysosomes.

Features and Benefits

This compound is featured on the PKA & PKG and PKB/Akt pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeremy H Herskowitz et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(49), 19086-19098 (2013-12-07)
Alzheimer's disease (AD) is the leading cause of dementia and has no cure. Genetic, cell biological, and biochemical studies suggest that reducing amyloid-β (Aβ) production may serve as a rational therapeutic avenue to delay or prevent AD progression. Inhibition of
Yuanbiao Zhao et al.
Nature communications, 6, 8243-8243 (2015-09-12)
Direct reprogramming of fibroblasts into cardiomyocytes by forced expression of cardiomyogenic factors, GMT (GATA4, Mef2C, Tbx5) or GHMT (GATA4, Hand2, Mef2C, Tbx5), has recently been demonstrated, suggesting a novel therapeutic strategy for cardiac repair. However, current approaches are inefficient. Here

Articles

Our offers many products related to PKA & PKG for your research needs.

We offer many products related to PKB/Akt for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service