Skip to Content
Merck
All Photos(1)

Key Documents

SML0571

Sigma-Aldrich

SB-612111 hydrochloride

≥98% (HPLC)

Synonym(s):

(5S,7S)-7-[[4-(2,6-Dichlorophenyl)-1-piperidinyl]methyl]-6,7,8,9-tetrahydro-1-methyl-5H-benzocyclohepten-5-ol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$228.00
25 MG
$619.00

$228.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$228.00
25 MG
$619.00

About This Item

Empirical Formula (Hill Notation):
C24H29Cl2NO · HCl
CAS Number:
Molecular Weight:
454.86
UNSPSC Code:
12352200
NACRES:
NA.77

$228.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1c(c(ccc1)Cl)C2CCN(CC2)C[C@@H]3C[C@@H](c4c(c(ccc4)C)CC3)O.Cl

InChI

1S/C24H29Cl2NO.ClH/c1-16-4-2-5-20-19(16)9-8-17(14-23(20)28)15-27-12-10-18(11-13-27)24-21(25)6-3-7-22(24)26;/h2-7,17-18,23,28H,8-15H2,1H3;1H/t17-,23-;/m0./s1

InChI key

DCKUVQADDNMWEF-DQFHVVJASA-N

Biochem/physiol Actions

SB-612111 is a potent antagonist of the nociceptin receptor ORL-1. SB-612111 binds to ORL-1 with high affinity (Ki = 0.33 nM) but has very low affinity for classic opioid receptors. The compound SB-612111 antagonizes the anti-morphine effects of nociceptin and blocks nociceptin-induced thermal hyperalgesia.
SB-612111 is an opiate receptor-like orphan receptor (ORL-1).

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to opioid receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service