Skip to Content
Merck
All Photos(1)

Key Documents

SML0565

Sigma-Aldrich

MK-0343

≥98% (HPLC)

Synonym(s):

7-Cyclobutyl-3-(2,6-difluorophenyl)-6-[(1-methyl-1H-1,2,4-triazol-5-yl)methoxy]-1,2,4-triazolo[4,3-b]pyridazine, MRK-409

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$485.00

$485.00


Estimated to ship on01 June 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
25 MG
$485.00

About This Item

Empirical Formula (Hill Notation):
C19H17F2N7O
CAS Number:
Molecular Weight:
397.38
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$485.00


Estimated to ship on01 June 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Cn1ncnc1COc2nn3c(nnc3cc2C4CCC4)-c5c(F)cccc5F

InChI

1S/C19H17F2N7O/c1-27-16(22-10-23-27)9-29-19-12(11-4-2-5-11)8-15-24-25-18(28(15)26-19)17-13(20)6-3-7-14(17)21/h3,6-8,10-11H,2,4-5,9H2,1H3

InChI key

GOIFCXRIFSYPFG-UHFFFAOYSA-N

Biochem/physiol Actions

MK-0343 (MRK-409) is a partial agonist for GABAA receptors with comparable Ki values for α1, α2, α3 and α5 containing receptors (0.2 – 0.4 nM). The compound has the greatest agonist activity against α3 receptors, and is brain penatrant, and displays anxiolytic activity in rodent and primate models.
MK-0343 is a GABAA partial agonist.

Features and Benefits

This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to GABAA receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service