Skip to Content
Merck
All Photos(1)

Documents

SML0563

Sigma-Aldrich

Docosahexaenoyl ethanolamide

≥98% (HPLC), ethanol solution (5 mg/ml)

Synonym(s):

(4Z,7Z,10Z,13Z,16Z,19Z)-N-(2-Hydroxyethyl)docosa-4,7,10,13,16,19-hexaenamide, DEA, DHEA, N-(2-Hydroxyethyl)-4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenamide, N-Docosahexaenoylethanolamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H37NO2
CAS Number:
Molecular Weight:
371.56
UNSPSC Code:
12352211
NACRES:
NA.77

Assay

≥98% (HPLC)

form

ethanol solution (5 mg/ml)

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

colorless to yellow

shipped in

dry ice

storage temp.

−20°C

Biochem/physiol Actions

Docosahexaenoyl ethanolamide (DHEA) has the ability to block the proliferation of head and neck squamous cell carcinoma (HNSCC) cell.
Docosahexaenoyl ethanolamide (DHEA) is a lipid signaling molecule present in brain and retina at concentrations, similar to those for arachidonoyl ethanolamide. DHEA binds to the rat brain CB1 receptor with a Ki of 324 nM. It exhibits anti-inflammatory and organ protective activity. Studies show that docosahexaenoyl ethanolamide is the mediator of neurite growth and synaptogenesis in hippocampal neurons, resulting in enhanced synaptic activity.

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5-lipoxygenase mediates docosahexaenoyl ethanolamide and N-arachidonoyl-L-alanine-induced reactive oxygen species production and inhibition of proliferation of head and neck squamous cell carcinoma cells
Park SW, et al.
BMC Cancer, 16(1), 458-458 (2016)

Articles

We offer many products related to cannabinoid receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service