Skip to Content
Merck
All Photos(1)

Key Documents

SML0550

Sigma-Aldrich

Nestorone

≥97% (HPLC)

Synonym(s):

17-(Acetyloxy)-16-methylene-19-norpregn-4-ene-3,20-dione, 17-Hydroxy-16-methylene-19-norpregn-4-ene-3,20-dione acetate, Elcometrine, Nestoron, ST-1435, ST1435

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$84.90
50 MG
$343.00

$84.90


Estimated to ship on25 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
10 MG
$84.90
50 MG
$343.00

About This Item

Empirical Formula (Hill Notation):
C23H30O4
CAS Number:
Molecular Weight:
370.48
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$84.90


Estimated to ship on25 May 2025

New, lower price on this item!

Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(=O)O[C@]1(C(C)=O)C(=C)C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C

InChI

1S/C23H30O4/c1-13-11-21-20-7-5-16-12-17(26)6-8-18(16)19(20)9-10-22(21,4)23(13,14(2)24)27-15(3)25/h12,18-21H,1,5-11H2,2-4H3/t18-,19+,20+,21-,22-,23-/m0/s1

InChI key

CKFBRGLGTWAVLG-GOMYTPFNSA-N

Gene Information

human ... PGR(5241)

Related Categories

Biochem/physiol Actions

Nestorone is a potent progesterone receptor agonist with no androgenic, estrogenic, or glucocorticoid-like activities.
Nestorone is a potent progesterone receptor agonist with no androgenic, estrogenic, or glucocorticoid-like activities. Nestorone has been used as a female contraceptive, and recent studies inidicate that it may be useful as a male contraceptive as well. It has also been shown to have neurogenic and neuroprotective activity.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vahid Mahabadi et al.
The Journal of clinical endocrinology and metabolism, 94(7), 2313-2320 (2009-04-16)
Testosterone (T) plus progestin combinations are the most promising hormonal male contraceptives. Nestorone (NES), a progestin without estrogenic or androgenic activity, when combined with T may be an excellent candidate for male contraception. Our objective was to determine the effect
Irving Sivin et al.
Contraception, 69(2), 137-144 (2004-02-05)
A 2-year trial of a single Nestorone (NES) rod implant was conducted at three Latin American centers, each enrolling 100 women. We studied the safety, effectiveness and acceptability of this progestin-releasing contraceptive implant. Three pregnancies occurred, the last at 18
Regine Sitruk-Ware
Human reproduction update, 12(2), 169-178 (2005-11-18)
The progestins have different pharmacologic properties depending upon the parent molecule, usually testosterone or progesterone (P), from which they are derived. Very small structural changes in the parent molecule may induce considerable differences in the activity of the derivative. In
Edith Weisberg et al.
Contraception, 72(1), 46-52 (2005-06-21)
We examined the clinical performance of contraceptive vaginal rings (rings) delivering Nestorone (NES) progestin and ethinyl estradiol (EE). Ring removal times were signaled by menstrual events. Bleeding patterns, adverse events, patterns of use and continuation rates were the principal parameters
Régine L Sitruk-Ware et al.
Contraception, 75(6), 430-437 (2007-05-24)
We evaluated the effects of a new combined hormonal contraceptive vaginal ring (CVR) delivering the nonandrogenic progestin Nestorone (NES) and ethinyl estradiol (EE) on several key estrogen-sensitive hepatic proteins that may be markers for the risk of arterial or venous

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service