Skip to Content
Merck
All Photos(1)

Key Documents

SML0515

Sigma-Aldrich

DCAP

≥95% (HPLC)

Synonym(s):

2-((3-(3,6-dichloro-9H-carbazol-9-yl)-2-hydroxypropyl)amino)-2-(hydroxymethyl)propane-1,3-diol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$295.00

$295.00


Estimated to ship on09 April 2025

New, lower price on this item!


Select a Size

Change View
25 MG
$295.00

About This Item

Empirical Formula (Hill Notation):
C19H22Cl2N2O4
CAS Number:
Molecular Weight:
413.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$295.00


Estimated to ship on09 April 2025

New, lower price on this item!

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

OCC(CO)(CO)NCC(O)Cn1c2ccc(Cl)cc2c3cc(Cl)ccc13

InChI

1S/C19H22Cl2N2O4/c20-12-1-3-17-15(5-12)16-6-13(21)2-4-18(16)23(17)8-14(27)7-22-19(9-24,10-25)11-26/h1-6,14,22,24-27H,7-11H2

InChI key

GETQKLUOZMYHGE-UHFFFAOYSA-N

Biochem/physiol Actions

DCAP is a potent broad-spectrum antibiotic that specifically targets the membranes of both Gram-positive and Gram-negative bacteria. It reduces the transmembrane potential, kills stationary-phase cells and sterilizes bacterial biofilms. DCAP has no effect on red blood cell membranes at levels toxic to bacteria.
DCAP is a potent broad-spectrum antibiotic.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service