Skip to Content
Merck
All Photos(1)

Key Documents

SML0513

Sigma-Aldrich

SKF83822 hydrobromide

Synonym(s):

6-Chloro-2,3,4,5-tetrahydro-1-(3-methylphenyl)-3-(2-propen-1-yl)-1H-3-Benzazepine-7,8-diol hydrobromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$111.00
25 MG
$437.00

$111.00


Estimated to ship on02 June 2025

New, lower price on this item!


Select a Size

Change View
5 MG
$111.00
25 MG
$437.00

About This Item

Empirical Formula (Hill Notation):
C20H22ClNO2 · HBr
CAS Number:
Molecular Weight:
424.76
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$111.00


Estimated to ship on02 June 2025

New, lower price on this item!

storage temp.

2-8°C

SMILES string

Br.Cc1cccc(c1)C2CN(CCc3c(Cl)c(O)c(O)cc23)CC=C

InChI

1S/C20H22ClNO2.BrH/c1-3-8-22-9-7-15-16(11-18(23)20(24)19(15)21)17(12-22)14-6-4-5-13(2)10-14;/h3-6,10-11,17,23-24H,1,7-9,12H2,2H3;1H

InChI key

CFWPKYBBXBANLU-UHFFFAOYSA-N

Biochem/physiol Actions

SKF83822 is a dopamine receptor agonist that only activates the D1 monomers and stimulates adenylate cyclase, but not PI hydrolysis.
SKF83822 is a dopamine receptor agonist that only activates the D1 monomers.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuji Odagaki et al.
Journal of receptor and signal transduction research, 39(1), 9-17 (2019-06-22)
Although multiple roles of dopamine through D1-like (D1 and D5) and D2-like (D2, D3, and D4) receptors are initiated primarily through stimulation or inhibition of adenylyl cyclase via Gs/olf or Gi/o, respectively, there have been many reports indicating diverse signaling

Articles

We offer many products related to dopamine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service