Skip to Content
Merck
All Photos(1)

Key Documents

SML0481

Sigma-Aldrich

Qc1

≥98% (HPLC)

Synonym(s):

1,2,3,4-Tetrahydro-4-oxo-N-(phenylmethyl)-2-thioxo-3-[3-(trifluoromethyl)phenyl]-7-quinazolinecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$115.00

$115.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$115.00

About This Item

Empirical Formula (Hill Notation):
C23H16F3N3O2S
CAS Number:
Molecular Weight:
455.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$115.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution, warmed)

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cccc(c1)N2C(=S)Nc3cc(ccc3C2=O)C(=O)NCc4ccccc4

InChI

1S/C23H16F3N3O2S/c24-23(25,26)16-7-4-8-17(12-16)29-21(31)18-10-9-15(11-19(18)28-22(29)32)20(30)27-13-14-5-2-1-3-6-14/h1-12H,13H2,(H,27,30)(H,28,32)

InChI key

IFNVTSPDMUUAFY-UHFFFAOYSA-N

Biochem/physiol Actions

Qc1 is a reversible inhibitor of threonine dehydroxygenase (TDH).
Qc1 is a reversible inhibitor of threonine dehydroxygenase (TDH). The IC50 value for inhibition of TDH by Qc1 is 500 nM, with no detectable inhibition of other dehydroxygenase enzymes at concentrations up to 10 mM. Qc1 induces arrest and autophagy of mouse ES cells with an EC50 of 3 mM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaiyu Kubota et al.
Biochimica et biophysica acta. General subjects, 1865(1), 129757-129757 (2020-10-05)
Trophoblast stem (TS) cell renewal and differentiation are essential processes in placentation. Special AT-rich binding protein 1 (SATB1) is a key regulator of the TS cell stem state. In this study, we identified SATB1 downstream targets and investigated their actions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service