Skip to Content
Merck
All Photos(1)

Key Documents

SML0480

Sigma-Aldrich

Terreic Acid

from Aspergillus terreus, ≥98% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$106.00
5 MG
$428.00

$106.00


Estimated to ship on17 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
1 MG
$106.00
5 MG
$428.00

About This Item

Empirical Formula (Hill Notation):
C7H6O4
CAS Number:
Molecular Weight:
154.12
UNSPSC Code:
12352106
NACRES:
NA.77

$106.00


Estimated to ship on17 May 2025

New, lower price on this item!

Request a Bulk Order

biological source

Aspergillus terreus

Quality Level

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: 10 mg/mL
H2O: 2 mg/mL

storage temp.

−20°C

InChI

1S/C7H6O4/c1-2-3(8)5(10)7-6(11-7)4(2)9/h6-8H,1H3/t6-,7+/m1/s1

InChI key

ATFNSNUJZOYXFC-RQJHMYQMSA-N

Application

Terreic Acid has been used to study its effect on alcohol intake and blood alcohol levels (BALs) in High Drinking in the Dark (HDID-1) mice.[1] It has also been used as a supplement for culturing yeast progenitor cells to study its effect on yeast replicative lifespan.[2]

Biochem/physiol Actions

Terreic acid (TA) is a covalent inhibitor of the bacterial cell wall biosynthetic enzyme MurA.
Terreic acid (TA) is a covalent inhibitor of the bacterial cell wall biosynthetic enzyme MurA. The survival of most bacteria depends on the functionality of this cytosolic enzyme. The inactivation of MurA requires the presence of UDP-N-acetylglucosamine 1-Carboxyvinyl transferase, which catalyzes the first step in the biosynthesis of the bacterial cell wall. TA is more potent than the known MurA inhibitor Fosphomycin. TA was also found to severely inhibit the transcription of TNFα and IL-2. TA inhibits protein synthesis by blocking the formation of leucyl-tRNA in sensitive bacteria and inhibits the catalytic effect of Bruton′s tyrosine kinase (Btk), an important factor in B-cell and mast cell activation, by inducing conformational changes that prevent Protein Kinase C (PKC) from interacting with the Btk domain. Moreover, recent studies have demonstrated that TA exhibits a very high level of radical scavenging activity.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Preparation Note

Store the product sealed at -20 °C. Under these conditions the product is stable for at least 2 years. Solution of the product in water is not stable and should be prepared immediately before use. Solution of the product in DMSO is stable for at least 2 months at 4 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuko Kawakami et al.
Allergology international : official journal of the Japanese Society of Allergology, 56(4), 403-409 (2007-08-24)
Atopic dermatitis is a chronic or chronically relapsing, pruritic inflammatory skin disease. The incidence of atopic dermatitis has dramatically increased during the past three decades in industrialized countries. We attempted to develop an improved method to induce an animal model
A high-throughput screen for yeast replicative lifespan identifies lifespan-extending compounds
Sarnoski EA, et al.
Cell Reports, 21(9), 2639-2646 (2017)
H Yamamoto et al.
The Japanese journal of antibiotics, 33(3), 320-328 (1980-03-01)
It was found that Aspergillus sp. No. Y-8980 which was isolated from a soil sample collected at Yoron Island in Kagoshima Prefecture belonged to Aspergillus terreus group by morphological observation. The active substance produced by the strain was obtained with
Genome-wide expression profiles drive discovery of novel compounds that reduce binge drinking in mice
Ferguson LB, et al.
Neuropsychopharmacology, 43(6) (2018)
T Subramanian et al.
Toxicology letters, 10(2-3), 249-253 (1982-02-01)
The binding of terreic acid to macromolecules such as nucleic acids and protein was examined. In vivo studies using [14C]terreic acid showed that radioactivity was incorporated into the protein and nucleic acid fractions of the liver mice. In vitro experiments

Articles

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service