Skip to Content
Merck
All Photos(1)

Documents

SML0438

Sigma-Aldrich

Mibolerone

≥98% (HPLC)

Synonym(s):

(7α,17β)-17-Hydroxy-7,17-dimethylestr-4-en-3-one, 7α,17α-Dimethyl-19-nortestosterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
EC Number:
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C20H30O2/c1-12-10-13-11-14(21)4-5-15(13)16-6-8-19(2)17(18(12)16)7-9-20(19,3)22/h11-12,15-18,22H,4-10H2,1-3H3/t12-,15+,16-,17+,18-,19+,20+/m1/s1

InChI key

PTQMMNYJKCSPET-OMHQDGTGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The synthetic androgen mibolerone (7α,17α dimethyl-19-nortestosterone) is an androgenic steroid.

Biochem/physiol Actions

Mibolerone is a synthetic anabolic steroid with a similar profile of activity as R1881 (Metribolone) with a higher affinity (Kd = 1.5 nM) for the androgen receptor in human prostate tissue than R1881 (Kd = 2.3 nM).
The synthetic androgen mibolerone (7α,17α dimethyl-19-nortestosterone) reversibly blocks the multiplication of LNCaP (human prostate carcinoma cell line) cells. In prostate, liver, and cultured cells, it serves as an efficient radioactive ligand for the quantitation and characterization of androgen receptors.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of radioactive 7α,17α?dimethyl?19?nortestosterone (mibolerone) in the assay of androgen receptors.
Schilling K and Liao S
Prostate, 5(6), 581-588 (1984)
Wayne Balkan et al.
Biochemical and biophysical research communications, 328(3), 783-789 (2005-02-08)
Despite their clinical importance for skeletal growth and homeostasis, the actions of androgens on osteoblastic cells are not well understood. MC3T3-E1 cells, a nontransformed murine preosteoblastic cell line, that traverse the stages of osteoblastic differentiation within 30 days in vitro
Saunders Handbook of Veterinary Drugs: Small and Large Animal (2015)
Eugene Bratoeff et al.
The Journal of steroid biochemistry and molecular biology, 107(1-2), 48-56 (2007-07-17)
In order to study the biological activity of the two novel steroidal carbamates derivatives: 8a and 8b, we determined the concentration of both compounds that inhibit the 50% of the activity of human prostate 5alpha-reductase enzyme, as well as the
Elisa J Cops et al.
The Journal of steroid biochemistry and molecular biology, 110(3-5), 236-243 (2008-06-03)
Androgen signaling, mediated by the androgen receptor (AR), is a critical factor influencing growth of normal and malignant breast cells. Given the increasing use of exogenous androgens in women, a better understanding of androgen action in the breast is essential.

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service