Skip to Content
Merck
All Photos(1)

Key Documents

SML0418

Sigma-Aldrich

ML-161

≥98% (HPLC)

Synonym(s):

2-Bromo-N-[3-[(1-oxobutyl)amino]phenyl]-benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$85.90
25 MG
$341.00

$85.90


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$85.90
25 MG
$341.00

About This Item

Empirical Formula (Hill Notation):
C17H17BrN2O2
CAS Number:
Molecular Weight:
361.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$85.90


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

CCCC(=O)Nc1cccc(NC(=O)c2ccccc2Br)c1

InChI

1S/C17H17BrN2O2/c1-2-6-16(21)19-12-7-5-8-13(11-12)20-17(22)14-9-3-4-10-15(14)18/h3-5,7-11H,2,6H2,1H3,(H,19,21)(H,20,22)

InChI key

DFOVLSMXPWPCFH-UHFFFAOYSA-N

Biochem/physiol Actions

ML-161 is an antagonist of the protease-activated receptor 1 (PAR1), that inhibits thrombin and SFLLN-induced platelet activation.
ML-161 is an antagonist of the protease-activated receptor 1 (PAR1), that inhibits thrombin and SFLLN-induced platelet activation. ML-161 dose dependently inhibits SFLLN-induced expression of P-selectin with an IC50 of 0.26 μM.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service