Skip to Content
Merck
All Photos(1)

Documents

SML0400

Sigma-Aldrich

ML204

≥98% (HPLC)

Synonym(s):

4-Methyl-2-(1-piperidinyl)-quinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N2
CAS Number:
Molecular Weight:
226.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL at warmed (clear solution)

storage temp.

2-8°C

SMILES string

Cc1cc(nc2ccccc12)N3CCCCC3

InChI

1S/C15H18N2/c1-12-11-15(17-9-5-2-6-10-17)16-14-8-4-3-7-13(12)14/h3-4,7-8,11H,2,5-6,9-10H2,1H3

InChI key

USYRQXDHKXGTCK-UHFFFAOYSA-N

Application

4-methyl-2-(1- piperidinyl)-quinoline (ML204) has been used to attenuate Ewing sarcoma (EWS)-FLI1 activity in TC32 cells. It has also been used to attenuate scratching behavior evoked by α-Me-5HT in mice.

Biochem/physiol Actions

ML204 is a potent and selective TRPC4 channel inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruizhi Zhang et al.
Polymers, 12(2) (2020-02-08)
In this study, we fabricated poly (methyl methacrylate) (PMMA) microcellular foams featuring tunable cellular structures and porosity, through adjusting the supercritical CO2 foaming conditions. Experimental testing and finite element model (FEM) simulations were conducted to systematically elucidate the influence of
Yun Zhu et al.
Chemico-biological interactions, 260, 84-90 (2016-11-08)
Aflatoxins are fungal metabolites which pose a major threat to food safety. Although these mycotoxins are established hepatocarcinogens, their effect on the reproductive organ is unknown. Transient Receptor Potential Channels (TRPs) are ubiquitously expressed in human tissues, including the placenta.
Englerin A inhibits EWS-FLI1 DNA binding in Ewing sarcoma cells
Caropreso V, et al.
The Journal of Biological Chemistry, 291(19), 10058-10066 (2016)
H M-H Chin et al.
Journal of helminthology, 91(4), 470-478 (2016-06-11)
Terrestrial arthropods host a variety of helminth parasites, yet quantifying the intensity of infection in these hosts post-mortem is challenging because carcasses may desiccate quickly. We recovered cysticercoids of Hymenolepis diminuta from desiccated flour beetle (Tribolium confusum) carcasses by modifying
Laura Bernal et al.
Science advances, 7(13) (2021-03-28)
Teeth are composed of many tissues, covered by an inflexible and obdurate enamel. Unlike most other tissues, teeth become extremely cold sensitive when inflamed. The mechanisms of this cold sensation are not understood. Here, we clarify the molecular and cellular

Articles

We offer many products related to transient receptor potential channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service