Skip to Content
Merck
All Photos(1)

Key Documents

SML0334

Sigma-Aldrich

DMPQ dihydrochloride

≥98% (HPLC)

Synonym(s):

5,7-Dimethoxy-3-(4-pyridinyl)quinoline dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$90.80

$90.80


Estimated to ship on20 March 2025

New, lower price on this item!


Select a Size

Change View
5 MG
$90.80

About This Item

Empirical Formula (Hill Notation):
C16H14N2O2 · 2HCl
CAS Number:
Molecular Weight:
339.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$90.80


Estimated to ship on20 March 2025

New, lower price on this item!

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: >10 mg/mL

storage temp.

room temp

SMILES string

Cl.Cl.COc1cc(OC)c2cc(cnc2c1)-c3ccncc3

InChI

1S/C16H14N2O2.2ClH/c1-19-13-8-15-14(16(9-13)20-2)7-12(10-18-15)11-3-5-17-6-4-11;;/h3-10H,1-2H3;2*1H

InChI key

YBBAOKYVJCNJIV-UHFFFAOYSA-N

Biochem/physiol Actions

DMPQ is a potent selective inhibitor of human vascular beta-type platelet derived growth factor receptor tyrosine kinase (PDGFR?) with IC50 = 80 nM. It is > 100-fold selective over EGFR, erbB2, p56, protein kinase A and protein kinase C.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the PDGFR page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wafa Altalhi et al.
Biomaterials, 119, 23-32 (2016-12-19)
Cell-based tissue engineering is a potential treatment alternative for organ replacement. However, the lack of a robust vasculature, especially in the context of diseases such as diabetes, is a major hindrance to its success. Despite extensive research on the effects
Akshaya Srinivasan et al.
Biomaterials, 167, 153-167 (2018-03-24)
Mesenchymal stem cells (MSCs) have been isolated from various mesodermal and ectodermal tissues. While the phenotypic and functional heterogeneity of MSCs stemming from their developmental origins has been acknowledged, the genetic and environmental factors underpinning these differences are not well-understood.

Articles

We offers many products related to PDGFR for your research needs.

Discover Bioactive Small Molecules for Kinase Phosphatase Biology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service