Skip to Content
Merck
All Photos(1)

Documents

SML0226

Sigma-Aldrich

Pseudocantharidin C

≥98% (HPLC)

Synonym(s):

(4-Bromophenyl) ((3aR,4R,7S,7aS)-rel-4,7-Epoxy-1H-isoindol-2(3H,3aH,4H,5H,6H,7H,7aH)-yl)methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16BrNO2
CAS Number:
Molecular Weight:
322.20
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥15 mg/mL

storage temp.

2-8°C

Biochem/physiol Actions

Pseudocantharidin C stimulates PP2A activity up to two-fold without influencing PP1. PP2A activation by pseudocantharidin C changes phosphorylation of tra2-beta1 at position T33 and promotes the SMN2 pre-mRNA exon 7 inclusion.

Features and Benefits

This compound is featured on the Phosphoprotein Phosphatases (Serine/Threonine) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to phosphoprotein phosphatases (Serine/Threonine) for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service