Skip to Content
Merck
All Photos(1)

Key Documents

SML0213

Sigma-Aldrich

MC2050 hydrate

≥98% (HPLC)

Synonym(s):

2-[2-(4-(2-pyridyl)-1-piperazinyl)ethylsulfanyl]-3H-quinazolin-4-one dihydrochloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$176.00
25 MG
$1,110.00

$176.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$176.00
25 MG
$1,110.00

About This Item

Empirical Formula (Hill Notation):
C19H21N5OS·2HCl · xH2O
Molecular Weight:
440.39 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$176.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: ≥5 mg/mL at warmed to 60 °C

storage temp.

2-8°C

SMILES string

O.Cl.Cl.O=C1NC(SCCN2CCN(CC2)c3ccccn3)=Nc4ccccc14

InChI

1S/C19H21N5OS.2ClH.H2O/c25-18-15-5-1-2-6-16(15)21-19(22-18)26-14-13-23-9-11-24(12-10-23)17-7-3-4-8-20-17;;;/h1-8H,9-14H2,(H,21,22,25);2*1H;1H2

InChI key

LMZZPRHHBPFDGE-UHFFFAOYSA-N

Application

MC2050 hydrate may be used in cell signaling studies.

Biochem/physiol Actions

MC2050 is a PARP-1 inhibitor
MC2050 is a water-soluble PARP-1 inhibitor. The IC50 for inhibition of PARP-1 activity is 119 nM, compared to 1.8 μM for PARP-2. MC2050 inhibits apoptosis and blocks poly ADP-ribosylation of histone H1 in hydrogen peroxide treated SH-SY5Y neuroblastoma cells.

Features and Benefits

This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Agents reported to activate cellular caspases include chemotherapeutic drugs, TNF receptor agonists, and other enzymes. Inhibitors of apoptosis were the first identified endogenous caspase inhibitors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service