Skip to Content
Merck
All Photos(1)

Key Documents

SML0185

Sigma-Aldrich

NQDI-1

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13NO4
CAS Number:
Molecular Weight:
319.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

orange

solubility

DMSO: ≥5 mg/mL (warmed to 60° C)

storage temp.

2-8°C

SMILES string

CCOC(=O)C1=C2c3ccccc3C(=O)c4cccc(NC1=O)c24

InChI

1S/C19H13NO4/c1-2-24-19(23)16-15-10-6-3-4-7-11(10)17(21)12-8-5-9-13(14(12)15)20-18(16)22/h3-9H,2H2,1H3,(H,20,22)

InChI key

UFJGFNHRMPMALC-UHFFFAOYSA-N

Application

NQDI-1 was used to study LPS-induced activation and role of ASK1 and p38 in anti-inflammatory cytokine production by macrophages.

Biochem/physiol Actions

ASK1 inhibitor
NQDI-1 inhibits the activation of caspase-3 and prevents cell death by apoptosis. It is neuroprotective and prevents the ROS-induced apoptosis of the neurons.
NQDI-1 is an inhibitor of apoptosis signal-regulating kinase (ASK1), Ki = 500 nM. NQDI-1 shows little or no inhibitory activity against Aurora A or ROCK kinases, or several receptor tyrosine kinases.

Features and Benefits

This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

So Yeong Cheon et al.
Frontiers in molecular neuroscience, 10, 261-261 (2017-09-01)
Inflammation is implicated in ischemic stroke and is involved in abnormal homeostasis. Activation of the immune system leads to breakdown of the blood-brain barrier and, thereby, infiltration of immune cells into the brain. Upon cerebral ischemia, infiltrated macrophages and microglia
Youguang Luo et al.
Oncotarget, 7(46), 75155-75164 (2016-09-23)
Pancreatic cancer has an extremely grim prognosis, with an overall 5-year survival rate less than 5%, as a result of its rapid metastasis and late diagnosis. To combat this disease, it is crucial to better understand the molecular mechanisms that
Koji Nomura et al.
Neuroscience letters, 549, 163-167 (2013-06-08)
Reactive oxygen species (ROS) are believed to be involved in many forms of neurodegeneration, including ischaemic infarct damage and Alzheimer's disease. Despite the known involvement of p38 and JNK MAP kinases in mediating apoptosis and cell death in a variety
Xi Huang et al.
International journal of cancer, 146(7), 1963-1978 (2019-08-08)
E3 ubiquitin ligases primarily determine the substrate specificity of the ubiquitin-proteasome system and play an essential role in the resistance to bortezomib in multiple myeloma (MM). Neural precursor cell-expressed developmentally downregulated gene 4-1 (NEDD4-1, also known as NEDD4) is a
Aiko Tanaka et al.
Biochemical and biophysical research communications, 493(1), 468-473 (2017-09-09)
Free fatty acids not only play a role in cell membrane construction and energy production but also exert diverse cellular effects through receptor and non-receptor mechanisms. Moreover, epidemiological and clinical studies have so far suggested that polyunsaturated fatty acids (PUFAs)

Articles

The mitogen-activated protein kinase (MAPK) family consists of stress-activated (SAPK) and MAPKs. Learn about their network of signal transduction cascades that mediate cellular responses to a diverse range of stimuli.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service