Skip to Content
Merck
All Photos(1)

Documents

SML0128

Sigma-Aldrich

M30 dihydrochloride

≥98% (HPLC)

Synonym(s):

5-[N-methyl-N-propargylaminomethyl]-8-hydroxyquinoline dihydrochloride, M-30 dihydrochloride, VAR10300 dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N2O · 2HCl
CAS Number:
Molecular Weight:
299.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

green-yellow

solubility

DMSO, heptane and xylene: ≥21 mg/mL

storage temp.

2-8°C

SMILES string

Cl.Cl.CN(CC#C)Cc1ccc(O)c2ncccc12

InChI

1S/C14H14N2O.2ClH/c1-3-9-16(2)10-11-6-7-13(17)14-12(11)5-4-8-15-14;;/h1,4-8,17H,9-10H2,2H3;2*1H

InChI key

XJNICPXVRPOSSO-UHFFFAOYSA-N

Application

M30 dihydrochloride may be used in monamine oxidase-mediated cell signaling studies.

Biochem/physiol Actions

M30 is a site-activated iron chelator and monoamine oxidase (MAO) inhibitor; with neuroprotective and anti-apoptotic activity. It is a multifunctional non-toxic, brain permeable iron chelator possessing a neuroprotective propargylamine moiety and an antioxidant-iron chelator moiety. M30 is a selective irreversible inhibitor of mitochondrial brain monoamine oxidases A and B (MAO-A and -B), providing for improved levels of neurotransmitters. It reduces oxidative damage and shows neuroprotective and neurorestorative effects in various neurodegenerative models of ALS, Alzheimer′s and Parkinson′s diseases, and is being investigated in other diseases where oxidative stress is a problem.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism and Histamine Synthesis and Metabolism pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Our company offers many products related to histamine synthesis and metabolism for your research needs.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service