Skip to Content
Merck
All Photos(1)

Key Documents

SML0077

Sigma-Aldrich

VU591

≥98% (HPLC)

Synonym(s):

2,2′-(oxydimethanediyl)bis(5-nitro-1H-benzimidazole), 2,2′-(oxydimethanediyl)bis(6-nitro-1H-benzimidazole), 5-nitro-2-{[(6-nitro-1H-benzimidazol-2-yl)methoxy]methyl}-1H-benzimidazole

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$91.80
25 MG
$370.00

$91.80


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$91.80
25 MG
$370.00

About This Item

Empirical Formula (Hill Notation):
C16H12N6O5
CAS Number:
Molecular Weight:
368.30
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$91.80


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥12 mg/mL

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc2nc(COCc3nc4ccc(cc4[nH]3)[N+]([O-])=O)[nH]c2c1

InChI

1S/C16H12N6O5/c23-21(24)9-1-3-11-13(5-9)19-15(17-11)7-27-8-16-18-12-4-2-10(22(25)26)6-14(12)20-16/h1-6H,7-8H2,(H,17,19)(H,18,20)

InChI key

ZQPXNYLXYNRFNP-UHFFFAOYSA-N

Gene Information

human ... KCNJ1(3758)

Application

VU591 may be used in cell signaling studies.

Biochem/physiol Actions

ROMK (Kir 1.1) inhibitor
VU591 is a Potassium Inwardly-Rectifying Channel (KCNJ1 or ROMK) specific inhibitor (IC50 = 240 nM) that is closely related to VU590. Unlike VU590, VU591 does not inhibit Kir7.1. The compound has a modest effect on Kir6.2/SUR1, causing 17% inhibition at 10 uM.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiun-Lin Horng et al.
Scientific reports, 7(1), 16215-16215 (2017-11-28)
Molecular mechanisms of Na
Guglielmo Schiano et al.
Scientific reports, 9(1), 19517-19517 (2019-12-22)
Uromodulin, the most abundant protein in normal urine, is produced by cells lining the thick ascending limb (TAL) of the loop of Henle. Uromodulin regulates the activity of the potassium channel ROMK in TAL cells. Common variants in KCNJ1, the

Articles

We offer many products related to potassium channels for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service