Skip to Content
Merck
All Photos(1)

Key Documents

SML0016

Sigma-Aldrich

CCG-63802

Synonym(s):

α-[[9-Methyl-2-(3-methylphenoxy)-4-oxo-4H-pyrido[1,2-a]pyrimidin-3-yl]methylene]-2-benzothiazoleacetonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$79.90
25 MG
$325.00

$79.90


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$79.90
25 MG
$325.00

About This Item

Empirical Formula (Hill Notation):
C26H18N4O2S
CAS Number:
Molecular Weight:
450.51
UNSPSC Code:
12352200
NACRES:
NA.77

$79.90


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

color

yellow

solubility

DMSO: ≥5 mg/mL (;warmed DMSO)

storage temp.

room temp

Application

CCG-63802 may be used to study G-protein-mediated cell signaling.

Biochem/physiol Actions

CCG-63802 is a reversible, allosteric inhibitor of RGS4
GCC-63802 binds to RGS4 and blocks interaction with Goa (IC50 = 1.9 uM). RGS4 (Regulator of G protein Signaling) is one of the RGP proteins responsible for terminating mu opioid receptor signaling. RSG proteins are GTPase activators, and drive G protein subunits to an inactive, GDP-bound form.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the G Proteins (Heterotrimeric) and Opioid Receptors pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Heterotrimeric G proteins modulate intracellular signals from 7TM receptors, impacting various cellular functions and drug targets.

We offers many products related to opioid receptors for your research needs.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service