Skip to Content
Merck
All Photos(1)

Key Documents

SMB01077

Sigma-Aldrich

Poliothyrsoside

≥85% (LC/MS-ELSD)

Synonym(s):

Nigracin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$584.00

$584.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$584.00

About This Item

Empirical Formula (Hill Notation):
C20H22O9
CAS Number:
Molecular Weight:
406.38
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

$584.00


Estimated to ship on16 April 2025


Request a Bulk Order

biological source

plant

Assay

≥85% (LC/MS-ELSD)

form

solid

mol wt

406.38

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1COC(=O)c3ccccc3)O)O)O)Oc2c(cc(cc2)O)CO

InChI

1S/C20H22O9/c21-9-12-8-13(22)6-7-14(12)28-20-18(25)17(24)16(23)15(29-20)10-27-19(26)11-4-2-1-3-5-11/h1-8,15-18,20-25H,9-10H2/t15-,16-,17+,18-,20-/m1/s1

InChI key

FLROYCKIIJCTDY-BFMVXSJESA-N

General description

Poliothyrsoside, also known as Nigracin, is a phenolic glycoside compound commonly found in plants such as Flacourtia indica, Loxocalyx urticifolius, Hydnocarpus annamensis, Xylosma flexuosa, Poliothyrsis sinensis, and Scolopia spinosa. Existing research suggests that this plant-derived metabolite exerts various biological activities, antioxidant, antidiabetic, anti-alzheimers and antiplasmodial properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical and biochemical research.

Biochem/physiol Actions

Poliothyrsoside is a pyruvic transaminase inhibitor known for its insulin-sensitizing effects in cell culture. Additionally, it exhibits anti-diabetic properties by inhibiting acetyl-CoA carboxylase, ultimately enhancing insulin sensitivity and lowering blood glucose levels in diabetic mice. This compound also serves as a lactate dehydrogenase inhibitor and demonstrates antidepressant-like actions, along with effectiveness against Alzheimer′s disease. In vitro studies have further revealed Poliothyrsoside′s capacity to inhibit virus production and exhibit antiviral activity against HIV-1.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gianluca Sferrazza et al.
Frontiers in pharmacology, 10, 1541-1541 (2020-02-11)
Drypetes klainei Pierre ex Pax is used in Cameroon by Baka people in the wound healing process and for the treatment of burns. In a previous paper we demonstrated the ability of both water (WE) and defatted methanol (DME) extracts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service