Skip to Content
Merck
All Photos(1)

Key Documents

SMB01063

Sigma-Aldrich

Kadsurin A

≥90% (LC/MS-ELSD)

Synonym(s):

(2S,3R,3aS,7aR)-2-(1,3-benzodioxol-5-yl)-3a,7a-dimethoxy-3-methyl-5-prop-2-enyl-3,7-dihydro-2H-1-benzofuran-6-one

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$466.00

$466.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$466.00

About This Item

Empirical Formula (Hill Notation):
C21H24O6
CAS Number:
Molecular Weight:
372.41
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

$466.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

372.41

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@]2([C@@]([C@@H]([C@H]1c3cc4c(cc3)OCO4)C)(C=C(C(=O)C2)CC=C)OC)OC

InChI

1S/C21H24O6/c1-5-6-15-10-20(23-3)13(2)19(27-21(20,24-4)11-16(15)22)14-7-8-17-18(9-14)26-12-25-17/h5,7-10,13,19H,1,6,11-12H2,2-4H3/t13-,19+,20+,21-/m1/s1

InChI key

YVRYZXAHRGGELT-MZNUGIIHSA-N

General description

Kadsurin A, a furanoid lignan, is a bioacive small molecule commonly found in stems of Kadsura coccinea. This compound exhibits potent biological activity, including anti-inflammatory, anti-paltelet, anti-cancer, and anti-viral properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

  • Exhibits potent anti-inflammatory properties by inhibiting pro-inflammatory cytokine release from macrophages.
  • Exerts protective effects against muscle injury in vivo and in vitro by effectively suppressing protease and cathepsin activity responsible for muscle tissue degradation.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service