Skip to Content
Merck
All Photos(1)

Key Documents

SMB01050

Sigma-Aldrich

Licochalcone B

≥85% (LC/MS-ELSD)

Synonym(s):

(E)-3-(3,4-dihydroxy-2-methoxyphenyl)-1-(4-hydroxyphenyl)prop-2-en-1-one, 3,4,4′-trihydroxy-2-methoxychalcone

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$577.00

$577.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$577.00

About This Item

Empirical Formula (Hill Notation):
C16H14O5
CAS Number:
Molecular Weight:
286.28
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

$577.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥85% (LC/MS-ELSD)

form

solid

mol wt

286.28

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O(C)c1c(c(ccc1\C=C\C(=O)c2ccc(cc2)O)O)O

InChI

1S/C16H14O5/c1-21-16-11(5-9-14(19)15(16)20)4-8-13(18)10-2-6-12(17)7-3-10/h2-9,17,19-20H,1H3/b8-4+

InChI key

DRDRYGIIYOPBBZ-XBXARRHUSA-N

General description

Licochalcone B is a prenylated flavonoid classified within the group of retrochalcones. This bioactive natural compound is commonly sourced from Glycyrrhiza species (G. glabra, G. uralensis, G. inflata, and G. aspera) plants. Existing research indicates that this plant-derived metabolite possesses a wide array of biological activities, including antibacterial, cardioprotective, anticancer, antioxidant, antiapoptotic, neuroprotective and anti-inflammatory properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

According to the existing research, Licochalcone B exerts its effects through multiple pathways, making it a versatile candidate for various therapeutic applications. It effectively combats inflammation by inhibiting NF-κB activation induced by LPS, leading to reduced production of NO, TNFα, and MCP-1. For bladder cancer, Licochalcone B demonstrates its potential by inducing S phase arrest, modulating cell cycle-related proteins, suppressing anti-apoptotic factors, and activating caspase-3. These actions collectively limit tumor growth in vivo, suggesting its utility in both bladder cancer therapy and prevention. Furthermore, Licochalcone B showcases robust antioxidant capabilities, successfully mitigating lipid peroxidation and lowering ROS production. It also exhibits anti-inflammatory properties by decreasing the release of NO, IL-6, and PGE2 in response to LPS. Furthermore, Licochalcone B emerges as a promising candidate for Alzheimer′s disease treatment, effectively preventing Aβ42 aggregation, chelating metal ions, and providing neuroprotection.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ze-Feng Wang et al.
Current medicinal chemistry, 27(12), 1997-2011 (2018-10-03)
Plants have always been an important source of medicines for humans, and licorice is a very significant herb in the development of humans. As a traditional herb, it is widely cultivated in China, Japan, Russia, Spain and India. With the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service