Skip to Content
Merck
All Photos(1)

Key Documents

SMB01025

Sigma-Aldrich

Alcesefoliside

≥90% (LC/MS-ELSD)

Synonym(s):

Quercetin 3-O-α-rhamnopyranosyl(1→6)-α-rhamnopyranosyl-(1→2)-β-galactopyranoside, Quercetin 3-O-(2,6-di-O-rhamnosyl)galactoside

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$584.00

$584.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$584.00

About This Item

Empirical Formula (Hill Notation):
C33H40O20
CAS Number:
Molecular Weight:
756.66
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

$584.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

756.66

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@@H]([C@H]([C@@H]1C)O)O)O)O[C@H]2[C@@H](O[C@@H]([C@@H]([C@@H]2O)O)CO[C@@H]6O[C@H]([C@@H]([C@H]([C@H]6O)O)O)C)OC3=C(Oc5c(c(cc(c5)O)O)C3=O)c4cc(c(cc4)O)O

InChI

1S/C33H40O20/c1-9-19(38)23(42)26(45)31(48-9)47-8-17-21(40)25(44)30(53-32-27(46)24(43)20(39)10(2)49-32)33(51-17)52-29-22(41)18-15(37)6-12(34)7-16(18)50-28(29)11-3-4-13(35)14(36)5-11/h3-7,9-10,17,19-21,23-27,30-40,42-46H,8H2,1-2H3/t9-,10-,17+,19-,20-,21-,23

InChI key

HKNBJSRIYRDSLB-MIORVHIISA-N

General description

Alcesefoliside is a flavonol tetraglycoside commonly available from Prunus mume, Monteverdia ilicifolia, Vicia amurensis, Chenopodium quinoa and Catharanthus roseus. Existing research suggests that this plant-derived secondary metabolite exerts various biological activities, including anti-inflammatory, antioxidant, antitumor and cytoprotective properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical and biochemical research.

Biochem/physiol Actions

  • Inhibits the production of pro-inflammatory cytokines and reactive oxygen species, resulting in anti-inflammatory and antioxidant effects.
  • Induces apoptosis in cancer cells, leading to antitumor effects.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service