Skip to Content
Merck
All Photos(1)

Key Documents

SMB01021

Sigma-Aldrich

Vitexin 4"-Ο-β-D-Glucoside

≥85% (LC/MS-ELSD)

Synonym(s):

4′′-O-Glucosylvitexin, Vitexin glucoside, Vitexin 4"-Ο-Glucoside

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$466.00

$466.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$466.00

About This Item

Empirical Formula (Hill Notation):
C27H30O15
CAS Number:
Molecular Weight:
594.52
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

$466.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥85% (LC/MS-ELSD)

form

solid

mol wt

594.52

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)c3c4[o]c(c[c](c4c(cc3O)O)=O)c5ccc(cc5)O)CO

InChI

1S/C27H30O15/c28-7-15-19(34)20(35)23(38)27(41-15)42-24-16(8-29)40-26(22(37)21(24)36)18-12(32)5-11(31)17-13(33)6-14(39-25(17)18)9-1-3-10(30)4-2-9/h1-6,15-16,19-24,26-32,34-38H,7-8H2/t15-,16-,19-,20+,21-,22-,23-,24-,26+,27+/m1/s1

InChI key

NDSUKTASTPEKBX-LXXMDOISSA-N

General description

Vitexin 4"-Ο-Glucoside, a flavonoid glucoside, is commonly found in Crataegus pinnatifida, Grataegus pinnatifida, and Passiflora incarnata. It effectively inhibits glutamate, a neurotransmitter that can impact brain function. Additionally, it has demonstrated the ability to suppress the production of pro-inflammatory cytokines and mitigate oxidative stress. Vitexin 4"-Ο-Glucoside exhibits a wide range of biological activities, including anti-inflammatory, antioxidant, and neuroprotective effects. Because of these properties, it holds significance in research pertaining to neurodegenerative conditions like Alzheimer′s and Parkinson′s diseases.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical and pharmaceutical research.

Biochem/physiol Actions

Inhibits glutamate release in the brain by lowering plasma concentrations.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service