Skip to Content
Merck
All Photos(1)

Key Documents

SMB01013

Sigma-Aldrich

Methyl 2,5-Dihydroxycinnamate

≥90% (LC/MS-ELSD)

Synonym(s):

2,4-Dihydroxymethylcinnamate, Methyl grevillate

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$657.00

$657.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$657.00

About This Item

Empirical Formula (Hill Notation):
C10H10O4
CAS Number:
Molecular Weight:
194.18
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

$657.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

194.18

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O(C)C(=O)C=Cc1c(ccc(c1)O)O

InChI

1S/C10H10O4/c1-14-10(13)5-2-7-6-8(11)3-4-9(7)12/h2-6,11-12H,1H3

InChI key

BQCNSTFWSKOWMA-UHFFFAOYSA-N

General description

Methyl 2,5-dihydroxycinnamate, a hydroxycinnamic acid, is a bioactive natural compound commonly derived from plants like Azadirachta indica and Murraya paniculata. Current research indicates that this plant-derived metabolite may possess diverse biological activities, encompassing anti-inflammatory and antitumor properties. Furthermore, it serves as a human urinary metabolite, a geroprotector, and an inhibitor of EC 2.7.10.1 (receptor protein-tyrosine kinase). It is a cell-permeable analog of erbstatin, a natural substance found in Streptomyces, demonstrates inhibitory effects on the autophosphorylation of the epidermal growth factor receptor. Moreover, it acts as a competitive inhibitor of epidermal growth factor receptor-associated tyrosine kinase, effectively impeding v-abl tyrosine kinase activation. Additionally, Methyl 2,5-dihydroxycinnamate has been observed to induce apoptosis in mouse thymocytes and inhibit the G2/M phase of the cell cycle.

Application

Methyl 2,5-Dihydroxycinnamate is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service