Skip to Content
Merck
All Photos(1)

Key Documents

SMB01001

Sigma-Aldrich

Loliolide

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$629.00

$629.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$629.00

About This Item

Empirical Formula (Hill Notation):
C11H16O3
CAS Number:
Molecular Weight:
196.24
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

$629.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

196.24

solubility

water: soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@@]2(C[C@H](CC(C2=CC1=O)(C)C)O)C

InChI

1S/C11H16O3/c1-10(2)5-7(12)6-11(3)8(10)4-9(13)14-11/h4,7,12H,5-6H2,1-3H3/t7-,11+/m0/s1

InChI key

XEVQXKKKAVVSMW-WRWORJQWSA-N

General description

Loliolide, a monoterpene lactone and a member of benzofurans, is a bioactive natural compound commonly derived from plants such as Lolium perenne, Euphorbia hirta, Canscora alata, Saussurea medusa, Xanthium spinosum, Sida acuta and Artemisia ludoviciana. Current research suggests that this metabolite acts as an inhibitor and may exhibit diverse biological activities, including antifungal, antitumor, cytoprotective, antibiotic, antioxidant, antimalarial, and anticancer properties.
Natural product derived from plant source.

Application

Loliolide is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and drug discovery research.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antioxidant efficacy of (-)-loliolide isolated from Sargassum horneri against AAPH-induced oxidative damage in Vero cells and zebrafish models in vivo
Kim H S, et al
Journal of Applied Phycology, 32, 3341-3348 (2020)
Hyo Hyun Yang et al.
Archives of pharmacal research, 38(5), 876-884 (2014-07-06)
Cellular senescence influences tumor suppression and progress, tissue repair and regeneration, tissue and organismal aging, and age-related diseases. Aging intervention might be an advantageous target for prevention and treatment of diverse age-related diseases. In this study, we investigated whether (-)-loliolide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service