Skip to Content
Merck
All Photos(1)

Key Documents

SMB00575

Sigma-Aldrich

3′,6-Disinapoylsucrose

≥90% (LC/MS-UV)

Synonym(s):

DSS

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$773.00

$773.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$773.00

About This Item

Empirical Formula (Hill Notation):
C34H42O19
CAS Number:
Molecular Weight:
754.69
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$773.00


Check Cart for Availability

Request a Bulk Order

Assay

≥90% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC1=C(OC)C=C(/C=C/C(O[C@@H]2[C@@](O[C@@H]3[C@H](O)[C@@H](O)[C@H](O)[C@@H](COC(/C=C/C4=CC(OC)=C(O)C(OC)=C4)=O)O3)(CO)O[C@H](CO)[C@H]2O)=O)C=C1OC

InChI

1S/C34H42O19/c1-45-18-9-16(10-19(46-2)26(18)39)5-7-24(37)49-14-23-28(41)30(43)31(44)33(50-23)53-34(15-36)32(29(42)22(13-35)52-34)51-25(38)8-6-17-11-20(47-3)27(40)21(12-17)48-4/h5-12,22-23,28-33,35-36,39-44H,13-15H2,1-4H3/b7-5+,8-6+/t22-,23-,28-,29-,30+,31-,32+,33-,34+/m1/s1

InChI key

FHIJMQWMMZEFBL-OPSYHMPNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Biochem/physiol Actions

3,6′-Disinapoylsucrose is a major active component of traditional Chinese medicine. It has significant effects on neuroprotection and improving learning memory.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Chen et al.
Biomedical chromatography : BMC, 23(12), 1326-1332 (2009-06-12)
3,6'-Disinapoylsucrose (DSS), a major active component of traditional Chinese medicine Yuan-Zhi (the roots of Polygala tenuifolia), has significant effects for neuroprotection and improving learning memory. In order to explore the pharmacokinetic properties of DSS so as to further understand its
Ying Chen et al.
Planta medica, 79(15), 1434-1439 (2013-09-18)
3,6'-Disinapoylsucrose is a major active component of the herb Polygala tenuifolia which has long been used for relieving tranquilization, uneasiness of the mind, and improving learning and memory. Our previous study found that 3,6'-disinapoylsucrose had a very low oral bioavailability.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service