Skip to Content
Merck
All Photos(1)

Key Documents

SMB00229

Sigma-Aldrich

Scillaren A

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$655.00

$655.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$655.00

About This Item

Empirical Formula (Hill Notation):
C36H52O13
CAS Number:
Molecular Weight:
692.79
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$655.00


Check Cart for Availability

Request a Bulk Order

Assay

≥90% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C[C@@H]1O[C@@H](O[C@H]2CC[C@]3(C)[C@H]4CC[C@]5(C)[C@H](CC[C@]5(O)[C@@H]4CCC3=C2)C6=COC(=O)C=C6)[C@H](O)[C@H](O)[C@H]1O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O

InChI

1S/C36H52O13/c1-17-31(49-33-29(42)27(40)26(39)24(15-37)48-33)28(41)30(43)32(46-17)47-20-8-11-34(2)19(14-20)5-6-23-22(34)9-12-35(3)21(10-13-36(23,35)44)18-4-7-25(38)45-16-18/h4,7,14,16-17,20-24,26-33,37,39-44H,5-6,8-13,15H2,1-3H3/t17-,20-,21+,22-,23+,24+,26+,27-,28-,29+,30+,31-,32-,33-,34-,35+,36-/m0/s1

InChI key

NXJOCELNFPGKIV-ARHXXGKOSA-N

General description

Natural product derived from plant source.
Scillaren A is a cardiac glycoside (bufadienolide) found in Drimia coromandeliana and D. razii.[1]

Application

Scillaren A has been used as a standard in spectroscopic estimation and reversed-phase high-performance liquid chromatography (RP-HPLC) analysis of cardiac glycosides from Drimia species.[1]

Biochem/physiol Actions

Scillaren A might inhibit Na+/K+-ATPase.[2]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Densitometric determination of proscillaridin A, scillaren A and scilliroside].
M Ferrer et al.
Il Farmaco; edizione pratica, 35(1), 32-46 (1980-01-01)
Phytochemicals, antioxidants, estimation of cardiac glycoside (Scillaren A) and detection of major metabolites using LC-MS from Drimia species
Yadav PB, et al.
South African Journal of botony (2020)
Studies on bronchodilator and cardiac stimulant activities of Urginea indica
Bashir S, et al.
Bangladesh Journal of Pharmacology, 8(3), 249-254 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service