Skip to Content
Merck
All Photos(1)

Key Documents

SMB00193

Sigma-Aldrich

Nordentatin

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$789.00

$789.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$789.00

About This Item

Empirical Formula (Hill Notation):
C19H20O4
CAS Number:
Molecular Weight:
312.36
UNSPSC Code:
12352205
NACRES:
NA.25

$789.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1C(C=Cc2c1c3c(c(c2O)C(C)(C)C=C)OC(=O)C=C3)(C)C

InChI

1S/C19H20O4/c1-6-18(2,3)14-15(21)11-9-10-19(4,5)23-16(11)12-7-8-13(20)22-17(12)14/h6-10,21H,1H2,2-5H3

InChI key

FREXEHTVBRSRGJ-UHFFFAOYSA-N

General description

Natural product derived from plant source.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biologically active coumarins from Enkleia siamensis.
Z Y Luo et al.
Journal of natural products, 49(6), 1161-1162 (1986-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service