Skip to Content
Merck
All Photos(3)

Key Documents

SILAW

Sigma-Aldrich

Silicic acid

BioReagent, suitable for column chromatography, 200-400 mesh (38 - 75 μm)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$230.00
1 KG
$1,150.00

$230.00


Check Cart for Availability


Select a Size

Change View
100 G
$230.00
1 KG
$1,150.00

About This Item

Empirical Formula (Hill Notation):
H2O3Si
CAS Number:
Molecular Weight:
78.10
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.54

$230.00


Check Cart for Availability

product line

BioReagent

Assay

≥99.9% Si (as SiO2) basis

form

solid

impurities

≤0.2% % nonvolatile with HF

particle size

200-400 mesh (38 - 75 μm)

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.02%

cation traces

Fe: ≤0.01%
heavy metals (as Pb): ≤0.001%

suitability

suitable for column chromatography

SMILES string

O[Si](O)=O

InChI

1S/H2O3Si/c1-4(2)3/h1-2H

InChI key

IJKVHSBPTUYDLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Silicic acid is used in protein chromatography. Silicic acid has been used in a study to demonstrate the anti-biofilm potential of a glycolipid surfactant produced by a tropical marine strain of Serratia marcescens. Silicic acid has also been used to study the antioxidant activity of fresh and processed Jalapeño and Serrano peppers.

Preparation Note

Acid-washed.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S K Gupta et al.
Infection and immunity, 62(10), 4572-4579 (1994-10-01)
This study investigated which adhesins of Pseudomonas aeruginosa interact with the glycolipid asialo GM1, using solid-phase binding and thin-layer chromatography assays. Radioiodinated pili and flagella contaminated with lipopolysaccharide (LPS) bound to the glycolipid. When LPS was reduced to acceptable levels
Koki Nagayama et al.
The Journal of antimicrobial chemotherapy, 50(6), 889-893 (2002-12-04)
The bactericidal activity of phlorotannins from brown algae against food-borne pathogenic bacteria (25 strains), methicillin-resistant Staphylococcus aureus (MRSA) (nine strains) and Streptococcus pyogenes (one strain) was examined and compared with that of catechins. In addition, the effect of the oral
Emilio Alvarez-Parrilla et al.
Journal of agricultural and food chemistry, 59(1), 163-173 (2010-12-04)
In this research, total phenols, flavonoids, capsaicinoids, ascorbic acid, and antioxidant activity (ORAC, hydroxyl radical, DPPH, and TEAC assays) of fresh and processed (pickled and chipotle canned) Jalapeño and Serrano peppers were determined. All fresh and processed peppers contained capsaicin
Devendra H Dusane et al.
Biofouling, 27(6), 645-654 (2011-06-29)
A tropical marine bacterium isolated from the hard coral, Symphyllia sp. was identified as Serratia marcescens on the basis of morphological, biochemical and 16S rDNA analysis. The bacterium showed antimicrobial activity towards the pathogens Candida albicans and Pseudomonas aeruginosa and
Hiroaki Saito et al.
Journal of oleo science, 64(6), 603-616 (2015-06-02)
The lipid and fatty acid composition of the muscle of the wild Japanese freshwater eel, Anguilla japonica, was analyzed between the initial and terminal stages of spawning migration to clarify the relationship between lipid physiology and maturation. Triacylglycerols were the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service